Skip to Content
MilliporeSigma

PC630

Anti-CD3 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-CD3 antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

dilution

(Frozen Sections (1:20-1:100)
Paraffin Sections (1:20-1:100, heat pre-treatment required))

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Anti-CD3, rabbit polyclonal, recognizes CD3 present in early thymocytes in human and mouse. It is validated for immunohistochemistry in frozen and paraffin sections.
Purified rabbit polyclonal antibody. Recognizes CD3.
Recognizes CD3 present in early thymocytes.

Immunogen

Human and Mouse
a synthetic peptide corresponding to amino acids from human CD3

Application


Frozen Sections (1:20-1:100)
Paraffin Sections (1:20-1:100, heat pre-treatment required)

Physical form

In proprietary buffer.

Analysis Note

Positive Control
Human tonsil

Other Notes

By immunohistochemical staining of tonsil tissue, this antibody stains T cell membranes. Antibody should be titrated for optimal results in individual systems.
Meuer, S. and Resch, K. 1989. Immunol. Today10, S23.
Clevers, H., et al. 1988. Eur. J. Immunol.18, 705.
Campana, D., et al. 1987. J. Immunol.138, 648.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Highly Toxic (H)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



João Moura et al.
Scientific reports, 9(1), 5836-5836 (2019-04-11)
Treatment for chronic diabetic foot ulcers is limited by the inability to simultaneously address the excessive inflammation and impaired re-epithelization and remodeling. Impaired re-epithelization leads to significantly delayed wound closure and excessive inflammation causes tissue destruction, both enhancing wound pathogen
Mariusz Gajda et al.
Folia histochemica et cytobiologica, 46(2), 143-146 (2008-06-04)
This paper presents a simple and reliable method of triple immunofluorescence staining that allows simultaneous detection of various cell types present in atherosclerotic plaque of apolipoprotein E and LDL receptor-double knockout (apoE/LDLR -/-) mice. We used combined direct and indirect



Global Trade Item Number

SKUGTIN
PC630-100UL04055977207811