MilliporeSigma
  • Triple immunofluorescence labeling of atherosclerotic plaque components in apoE/LDLR -/- mice.
Triple immunofluorescence labeling of atherosclerotic plaque components in apoE/LDLR -/- mice.
Folia histochemica et cytobiologica (2008-06-04)
Mariusz Gajda, Jacek Jawień, Lukasz Mateuszuk, Grzegorz J Lis, Andrzej Radziszewski, Stefan Chłopicki, Jan A Litwin
ABSTRACT
This paper presents a simple and reliable method of triple immunofluorescence staining that allows simultaneous detection of various cell types present in atherosclerotic plaque of apolipoprotein E and LDL receptor-double knockout (apoE/LDLR -/-) mice. We used combined direct and indirect procedures applying commercially available primary antibodies raised in different species to detect smooth muscle cells (Cy3-conjugated mouse anti-smooth muscle actin, SMA), macrophages (rat anti-CD68) and T lymphocytes (rabbit anti-CD3). Fixation of the material in acetone and modified incubation protocol employing nonfat dry milk in preincubation and incubation media significantly increased the intensity of labeling and effectively quenched the background. Our method offers an efficient way to detect qualitative as well as quantitative changes of macrophages, T lymphocytes and smooth muscle cells in atherosclerotic plaque of apoE/LDLR -/- mice during atherosclerosis development or in response to pharmacological treatment.
Sigma-Aldrich
Anti-CD3 Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture