Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PBTK07610

Millipore

PES Membrane Filter, 30 kDa NMWCO

Biomax®, filter diam. 76 mm

Synonym(s):

Biomax PES ultrafiltration membrane discs, Ultrafiltration Discs, 30 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

material

polyethersulfone membrane

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Biomax®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

76 mm

filter diam.

76 mm

filtration area

41.8 cm2

pore size

30 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

  • Ultrafiltration cut disc membranes in Biomax® polyethersulfone (PES) are ideal for protein concentration, purification, and desalting.
  • Optimized for concentrating or desalting serum, plasma, or conditioned tissue culture media.
  • A wide range of pore sizes is available to suit your application needs.

Application

30 kDa NMWL, Biomax polyethersulfone, 76 mm diameter

Linkage

Replaces: 13242

Legal Information

BIOMAX is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service