Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

UFSC40001

Amicon® Stirred Cell 400mL

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Synonym(s):

Amicon stirred cell

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031690

material

Nylon assembly (cap and tube fitting)
Nylon stand (retaining)
Tygon® tube (filtrate)
anodized aluminum stand (retaining)
polyacetal assembly (stirring)
polyethylene tube (pressure)
polysulfone assembly (stirring)
polysulfone holder (cylinder & membrane)
silicone O-ring

Quality Level

product line

Amicon®

manufacturer/tradename

Amicon®

parameter

400 mL process volume

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

H

18.7 cm (7.4 in.)

filter diam.

76 mm

filtration area

41.8 cm2

shipped in

ambient

Application

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Principle

Pressure

Linkage

Replaces: 5124

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service