Skip to Content
MilliporeSigma

MXLMC0120

Milliflex® Liquid Media Cassette

For bioburden and water analysis with the Milliflex® system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41116129
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

styrene-acrylonitrile (SAN)

Quality Segment

agency

EP, USP

sterility

sterile

form

plate (liquid agar)

feature

ready-to-use

manufacturer/tradename

Milliflex®

input

sample type cosmetics
sample type beer
sample type pharmaceutical(s)
sample type bottled water

application(s)

bioburden testing
cosmetics
microbiology
pharmaceutical
water monitoring

shipped in

ambient

General description

Milliflex® Liquid Media Cassette is designed to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid, and reliable bioburden testing. The liquid media cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate can hold approximately 2mL of a liquid media dispensed by the user. Broth media in ampoule is typically used in combination with the Milliflex® Liquid Media Cassette (Ampouled liquid culture media for the Milliflex® system)..

Application

Milliflex® Liquid Media Cassette is used in Bioburden testing and general microbiological analysis of water. It is widely used in the cosmetic, and brewing industry.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library