Skip to Content
MilliporeSigma

MXSMEND48

Milliflex® Pre-filled Media Cassettes

m-Endo LES Agar, for coliforms, pack of 48 tests, suitable for bioburden testing

Synonym(s):

m-Endo LES Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116129
NACRES:
NA.74
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

USP International standards

Quality Segment

sterility

aseptically filled

form

plate (pre-poured agar)

shelf life

6 mo.

feature

ready-to-use

packaging

cassette of 10 mL, pack of 48 tests

manufacturer/tradename

Milliflex Oasis®

storage condition

protect from light (right side up)

color

slightly opalescent rose

input

sample type cosmetics
sample type beer
sample type pharmaceutical(s)
sample type bottled water

pH

7.2±0.2 (25.0 °C)

application(s)

bioburden testing, cosmetics
microbiology
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® System

shipped in

ambient

storage temp.

2-8°C

suitability

suitable for (suitable for solid cultivation), coliforms

General description

Milliflex® Cassette are ready to use agar-based media to be used with Milliflex® or Milliflex Oasis® system for convenient, rapid and reliable bioburden testing. The prefilled agar cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate is a 3-piece assembly containing approximately 10ml of the media. m-Endo LES Agar is a selective media for detecting total coliforms in drinking water. Sodium lauryl sulphate and sodium desoxycholate are inhibitors of gram-positive bacteria, basic Fuchsin is a pH indicator, sodium sulphite is added to decolorize the basic fuchsin solution and the ethanol additive increases the antibacterial nature of the formulation. Lactose fermenting bacteria produce acetaldehyde that reacts with the sodium sulphite and fuchsin to form red colonies. The development of a metallic sheen occurs when the organism produces aldehydes with the rapid fermentation of lactose. The sheen will be suppressed if the inoculum is too heavy.

Application

Milliflex® Cassette prefilled with m-Endo LES Agar is used in Bioburden testing and general microbiological analysis of water, wastewater and dairy products. It is widely used in pharmaceutical, cosmetic and brewing industry.

Preparation Note

m-Endo LES

Analysis Note

Coliform colonies appear deep red with distinct green metallic sheen
QC organisms
E. coli (ATCC 25922)

P. aeruginosa (ATCC 9027)

E. aerogenes (ATCC 49701)

S. aureus (ATCC 6538) as negative control
Target Microorganisms: Coliform species

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library