Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MMQUANK01

Millipore

Milliflex® Quantum Rapid Detection System

Supplied with 1 Reader, 1 Reader Stand, 1 Membrane Transfer Tool, 1 Removal Rack, 1 Camera and Installation flash drive

Synonym(s):

Fluorescence Based Microbial Detection System, Rapid Microbial Detection System

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

AC/DC input

100 V / 200 V, 50-60 Hz

material

304L stainless steel (Camera front plate)
304L stainless steel (Optical Chamber, Reader)
304L stainless steel handle (Reader)
304L stainless steel stand (Reader)
PVC foot (Reader)
aluminum foot (with epoxy coating, Camera)
aluminum housing (Sheet, Reader)
glass window(s) (Camera)
polyoxymethylene housing (Membrane Transfer Tool)
polyphenylsulfone (PPSU) (Removal Rack Membrane Transfer Tool )
silicone seal (Membrane Transfer Tool)

description

The Milliflex® Quantum Reader and Camera can be used under a Laminar Flow Hood

manufacturer/tradename

Milliflex®

parameter

4.4 kg operating weight
45 °C temperature (for samples)

W × D × H

5.6 in. × 9.8 in. × 4.9 in.

application(s)

pharmaceutical

compatibility

for use with Milliflex® Quantum system.

General description

Milliflex® Quantum Rapid Detection System is a rapid fluorescent-based solution designed for fast quantitative detection of microorganisms over a broad range of filterable matrices. The easy-to-use simple system uses industry standard membrane filtration techniques to detect viable and culturable microbes down to 1CFU per sample.
The Milliflex® Quantum Rapid Detection System not only delivers rapid test results, it also enables the user to continue to grow the microbes in order to identify them later.

Application

Milliflex® Quantum Rapid Detection System is used to quantify and detect microorganisms from filtered samples. The broad range of filterable matrices raw material (media, buffers, pharmaceutical ingredients and water), in process samples, final product and environmental samples.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

Milliflex® Quantum system enables rapid microbial contamination detection with fluorescence staining technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service