Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIX0010LG05

Millipore

Mobius® 10L Mixer Bag Assembly

Gold Mobius® 10L mixer bag

Synonym(s):

Mobius® 10 L mixer bag assy, Gold

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103801
eCl@ss:
32010600

Quality Level

product line

Mobius®

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service