Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIACTUB01

Millipore

Microfil® Silicon Tubing

With Tee and connections for use with Microfil® filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

silicone

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Microfil® with S-Pak®

input

sample type beverage(s)
sample type food(s)

application(s)

bioburden testing
clinical
environmental
food and beverages
industrial qc
water monitoring

compatibility

for use with Microfil® filtration system

shipped in

ambient

General description

The silicon tubing is an additional accessory of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. The tube is used to connect the pump to the manifold or filtration apparatus using the Tee connectors. The tubing is autoclavable, 5 m long, with an internal diameter of 9.5 mm.

Application

The silicon tubing is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing, and water monitoring.

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service