MilliporeSigma
  • Inhibition of Upf2-Dependent Nonsense-Mediated Decay Leads to Behavioral and Neurophysiological Abnormalities by Activating the Immune Response.
Inhibition of Upf2-Dependent Nonsense-Mediated Decay Leads to Behavioral and Neurophysiological Abnormalities by Activating the Immune Response.
Neuron (2019-10-06)
Jennifer L Johnson, Loredana Stoica, Yuwei Liu, Ping Jun Zhu, Abhisek Bhattacharya, Shelly A Buffington, Redwan Huq, N Tony Eissa, Ola Larsson, Bo T Porse, Deepti Domingo, Urwah Nawaz, Renee Carroll, Lachlan Jolly, Tom S Scerri, Hyung-Goo Kim, Amanda Brignell, Matthew J Coleman, Ruth Braden, Usha Kini, Victoria Jackson, Anne Baxter, Melanie Bahlo, Ingrid E Scheffer, David J Amor, Michael S Hildebrand, Penelope E Bonnen, Christine Beeton, Jozef Gecz, Angela T Morgan, Mauro Costa-Mattioli
ABSTRACT
In humans, disruption of nonsense-mediated decay (NMD) has been associated with neurodevelopmental disorders (NDDs) such as autism spectrum disorder and intellectual disability. However, the mechanism by which deficient NMD leads to neurodevelopmental dysfunction remains unknown, preventing development of targeted therapies. Here we identified novel protein-coding UPF2 (UP-Frameshift 2) variants in humans with NDD, including speech and language deficits. In parallel, we found that mice lacking Upf2 in the forebrain (Upf2 fb-KO mice) show impaired NMD, memory deficits, abnormal long-term potentiation (LTP), and social and communication deficits. Surprisingly, Upf2 fb-KO mice exhibit elevated expression of immune genes and brain inflammation. More importantly, treatment with two FDA-approved anti-inflammatory drugs reduced brain inflammation, restored LTP and long-term memory, and reversed social and communication deficits. Collectively, our findings indicate that impaired UPF2-dependent NMD leads to neurodevelopmental dysfunction and suggest that anti-inflammatory agents may prove effective for treatment of disorders with impaired NMD.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Premixed 32 Plex - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid