Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MCKP1-110K

Millipore

MILLIPLEX® Mouse Immuno-Oncology Checkpoint Protein Panel 1

Immuno-Oncology Multiplex Assay

Synonym(s):

luminex mouse immuno oncology checkpoint protein multiplex kit, millipore mouse IO checkpoint protein multiplex, mouse I O cancer immunoassay panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 101
(Galectin-1)

accuracy: 103
(CD80/B7-1)

accuracy: 103
(IFNγ)

accuracy: 104
(CTLA-4/CD152)

accuracy: 104
(PD-L2)

accuracy: 104
(TLR-2)

accuracy: 105
(4-1BBL/TNFSF9)

accuracy: 105
(BCA-1/CXCL-13)

accuracy: 105
(CD25/IL-2Rα)

accuracy: 105
(IL-10)

accuracy: 105
(LAG-3)

accuracy: 106
(5’-NT/CD73)

accuracy: 106
(PD-1)

accuracy: 106
(TIM-3)

accuracy: 106
(TNFα)

accuracy: 107
(CD137/4-1BB)

accuracy: 107
(CD27)

accuracy: 108
(E-Cadherin)

accuracy: 108
(Galectin-3)

accuracy: 108
(HVEM)

accuracy: 109
(CD40)

accuracy: 109
(GITR)

accuracy: 110
(B7-H2/ICOSL)

accuracy: 110
(CD226/DNAM-1)

accuracy: 111
(BTLA)

accuracy: 94
(GranzymeB)

accuracy: 95
(B7-H3/CD276)

accuracy: 96
(PD-L1)

standard curve range: 1.4 - 1000 pg/mL
(HVEM)

standard curve range: 11 - 8000 pg/mL
(IL-10)

standard curve range: 137-100,000 pg/mL
(5’-NT/CD73)

standard curve range: 14 - 10000 pg/mL
(CD226/DNAM-1)

standard curve range: 14 - 10000 pg/mL
(CD40)

standard curve range: 14 - 10000 pg/mL
(PD-L1)

standard curve range: 14-10,000 pg/mL
(4-1BBL/TNFSF9)

standard curve range: 2.1 - 1500 pg/mL
(IFNγ)

standard curve range: 2.7 - 2000 pg/mL
(CD25/IL-2Rα)

standard curve range: 21-15000 pg/mL
(BTLA)

standard curve range: 27-20000 pg/mL
(B7-H2/ICOSL)

standard curve range: 27-20000 pg/mL
(BCA-1/CXCL-13)

standard curve range: 3.4 - 2500 ng/mL
(Galectin-1)

standard curve range: 3.4-2500 ng/mL
(B7-H3/CD276)

standard curve range: 34.3 - 25000 pg/mL
(CD80/B7-1)

standard curve range: 342 - 250000 pg/mL
(E-Cadherin)

standard curve range: 4.1 - 3000 pg/mL
(TNFα)

standard curve range: 41 - 30000 pg/mL
(GITR)

standard curve range: 41 - 30000 pg/mL
(Galectin-3)

standard curve range: 412 - 300000 pg/mL
(LAG-3)

standard curve range: 412 - 300000 pg/mL
(TLR-2)

standard curve range: 5.5 - 4000 pg/mL
(CD137/4-1BB)

standard curve range: 5.5 - 4000 pg/mL
(CTLA-4/CD152)

standard curve range: 5.5 - 4000 pg/mL
(TIM-3)

standard curve range: 55 - 40000 pg/mL
(CD27)

standard curve range: 55 - 40000 pg/mL
(PD-1)

standard curve range: 6.9 - 5000 pg/mL
(PD-L2)

standard curve range: 8.2 - 6000 pg/mL
(GranzymeB)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Immune checkpoint proteins are key regulators of the immune system and drug targets of cancer immunotherapy. Immune checkpoint molecules have been found on the cell surface of immune cells and tumor cells as activating receptors or inhibitory receptors. Immune checkpoint inhibitors, such as blocking antibodies to cytotoxic T lymphocyte-associated antigen-4 (CTLA-4), programmed cell death protein-1 (PD-1) and programmed cell death ligand 1 (PD-L1), have demonstrated significant efficacy in the treatment of a variety of cancers. Recent studies have also shown many immune checkpoint proteins can undergo alternative splicing or proteolytic cleavage to produce soluble checkpoint proteins. The expression levels of soluble and cell-surface immune checkpoint proteins in the circulation and in tumor-microenvironment (TME) may be used as putative immune-based biomarkers for cancer research and biomarker discovery in immunological diseases.

MILLIPLEX® Mouse Immuno-Oncology Checkpoint Protein Panel 1 is a 28-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma, cell culture supernatant, cell lysate, and tissue/tumor extract samples: 4-1BBL/TNFSF9, 5′-NT/CD73, B7-H2/ICOSL, B7-H3/CD276, BCA-1/CXCL-13, BTLA, CD137/4-1BB, CD226/DNAM-1, CD25/IL-2Rα, CD27, CD40, CD80/B7-1, CTLA-4/CD152, E-cadherin, Galectin-1, Galectin-3, GITR, Granzyme B, HVEM, IFNγ, IL-10, LAG-3, PD-1, PD-L1, PD-L2, TIM-3, TLR-2, and TNFα.

Panel Type: MAGNETIC Circulating Cancer

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible

Application

The Mouse Immuno-Oncology Checkpoint Protein Panel 1 bead-based multiplex panel, using the Luminex® xMAP® technology, enables the simultaneous analysis of 28 immune checkpoint proteins in mouse serum, plasma, cell culture supernatant, cell lysate, and tissue/tumor extract samples.
This is an overnight incubation assay (16-18 hours at 2-8 °C).
A maximum of 25 μL per well 1:2 diluted serum or plasma can be used. Tissue culture samples may be run neat or appropriately diluted in control medium. Note: Best results using cell/tissue lysate samples may require diluting in assay buffer while testing similarly diluted lysis buffer for matrix effects.

Research Category: Cancer/Immuno-oncology

Analytes:
4-1BBL/TNFSF9
5′-NT/CD73
B7-H2/ICOSL
B7-H3/CD276
BCA-1/CXCL-13
BTLA
CD137/4-1BB
CD226/DNAM-1
CD25/IL-2Rα
CD27
CD40
CD80/B7-1
CTLA-4/CD152
E-cadherin
Galectin-1
Galectin-3
GITR
Granzyme B
HVEM
IFNγ
IL-10
LAG-3
PD-1
PD-L1
PD-L2
TIM-3
TLR-2
TNFα

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

96-well plate

Other Notes

Sensitivity: Refer to kit protocol for sensitivities for individual cytokines/chemokines.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® panel multiplexes immune checkpoint molecules, aiding NSCLC research to identify immunotherapy targets for PD-1/PD-L1 therapy.

MILLIPLEX® panel multiplexes immune checkpoint molecules, aiding NSCLC research to identify immunotherapy targets for PD-1/PD-L1 therapy.

MILLIPLEX® panel multiplexes immune checkpoint molecules, aiding NSCLC research to identify immunotherapy targets for PD-1/PD-L1 therapy.

MILLIPLEX® panel multiplexes immune checkpoint molecules, aiding NSCLC research to identify immunotherapy targets for PD-1/PD-L1 therapy.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service