Skip to Content
MilliporeSigma

MCE5001FS1

Millipore

Millistak+® Pod Depth Filter

CE50, cellulose, filtration area 0.11 m2, hydrophilic

Synonym(s):

Millistak+ Pod Depth Filter, CE50 media series, 0.11 m 2 surface area, 1/pk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

material

cellulose
glass filled polypropylene

Quality Level

description

For clarification of serum, plasma, vaccines, cell culture or other fluids.

feature

hydrophilic

manufacturer/tradename

Millistak+®

parameter

1.0 bar max. differential pressure (15 psid) at 80 °C (Forward)
1.0 bar max. inlet pressure (15 psid) at 80 °C
2.1 bar max. differential pressure (30 psid) at 25 °C (Forward)
2.1 bar max. differential pressure (30 psid) at 25 °C (Reverse)
3.5 bar max. inlet pressure (50 psid) at 25 °C

H

32 cm (12.5 in.)

L

62 cm (24.2 in.)

filtration area

0.11 m2

impurities

<0.25 EU/mL bacterial endotoxins (LAL test, Aqueous extraction)

input

sample type liquid

conductivity

1.52-1.94 μS/cm at 25 °C (product water)

filter grade

CE50

General description

The Millistak+® depth filter media is offered® in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MCE5001FS1 comes in a POD format featuring a 32 cm (H) x 62 cm (L) and a filtration area of 0.11 m2.

This CE50 media series delivers optimal performance through a gradient density matri, ideal for primary (coarse) clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.

The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.

Easy to Use

With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.

Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.

Application

For clarification of serum, plasma, vaccines, cell culture or other fluids.

Features and Benefits

  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Specifications

Filter Grade: CE50
Per 24-hour static soak in 10 L pure water post autoclave (1 cycle of 30 minutes at 123 °C) and pure water flush of 10 L per ft2 (100 L/m2) of surface area.
Sodium: ≤9.91 mg/ft2
Calcium: ≤1.46 mg/ft2
Magnesium: ≤0.662 mg/ft2
Mercury: ≤0.0004 mg/ft2
Chromium: 0.0003 mg/ft2
Aluminum: ≤0.059 mg/ft2
Potassium: ≤0.042 mg/ft2
Lead: ≤0.0004 mg/ft2
Arsenic: ≤0.002 mg/ft2
Cobalt: 0.0002 mg/ft2
Iron: ≤0.042 mg/ft2
Manganese: ≤0.001 mg/ft2
Titanium: <0.001 mg/ft2
Zinc: ≤0.003 mg/ft2
Nickel: ≤0.0004 mg/ft2
Copper: ≤0.0004 mg/ft2

Preparation Note

Sterilization Method
1 autoclave cycle of 60 minutes at 123 °C

Analysis Note

Product Water Conductivity at 25°C: post autoclave (1 cycle of 60 minutes at 123 °C) and pure water flush of 5 liters per ft2 (50 L/m2) of media surface area

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service