MilliporeSigma
  • Integrin β1 is a key determinant of the expression of angiotensin-converting enzyme 2 (ACE2) in the kidney epithelial cells.
Integrin β1 is a key determinant of the expression of angiotensin-converting enzyme 2 (ACE2) in the kidney epithelial cells.
European journal of cell biology (2023-04-22)
Md Saimon Mia, Delowar Hossain, Emerson Woodbury, Sean Kelleher, Riya J Palamuttam, Reena Rao, Preston Steen, Yagna Pr Jarajapu, Sijo Mathew
ABSTRACT
The expression of the angiotensin-converting enzyme 2 (ACE2) is altered in multiple chronic kidney diseases like hypertension and renal fibrosis, where the signaling from the basal membrane proteins is critical for the development and progression of the various pathologies. Integrins are heterodimeric cell surface receptors that have important roles in the progression of these chronic kidney diseases by altering various cell signaling pathways in response to changes in the basement membrane proteins. It is unclear whether integrin or integrin-mediated signaling affects the ACE2 expression in the kidney. The current study tests the hypothesis that integrin β1 regulates the expression of ACE2 in kidney epithelial cells. The role of integrin β1 in ACE2 expression in renal epithelial cells was investigated by shRNA-mediated knockdown and pharmacological inhibition. In vivo studies were carried out using epithelial cell-specific deletion of integrin β1 in the kidneys. Deletion of integrin β1 from the mouse renal epithelial cells reduced the expression of ACE2 in the kidney. Furthermore, the downregulation of integrin β1 using shRNA decreased ACE2 expression in human renal epithelial cells. ACE2 expression levels were also decreased in renal epithelial cells and cancer cells when treated with an integrin α2β1 antagonist, BTT 3033. SARS-CoV-2 viral entry to human renal epithelial cells and cancer cells was also inhibited by BTT 3033. This study demonstrates that integrin β1 positively regulates the expression of ACE2, which is required for the entry of SARS-CoV-2 into kidney cells.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 12G10, clone 12G10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin beta1 (CD29) Antibody, clone mAb13, clone mAb13, from rat