Skip to Content
MilliporeSigma

P2PLBCC05

Pellicon® 2 Cassette Media: 3 kDa

Chemistry: Regenerated Cellulose (RC) Screen Type: C Device Size: 0.5 m2

Synonym(s):

Pellicon® 2 Cassette Ultrafiltration Module PLBC C 0.5 m2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32031623
UNSPSC Code:
23151806
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pellicon® 2 Cassette Media: 3 kDa, Chemistry: Regenerated Cellulose (RC) Screen Type: C Device Size: 0.5 m2

Quality Segment

product line

Pellicon®

technique(s)

analytical sample preparation: suitable, protein purification: suitable

H

1.5 cm (0.6 in.)

L

21 cm (8.3 in.)

Legal Information

Pellicon is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Pellicon® 2 Cassette Media: 3 kDa


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library