Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS528

Sigma-Aldrich

Anti-HGPRT Antibody, clone 13H11.1

clone 13H11.1, from mouse

Synonym(s):

Hypoxanthine-guanine phosphoribosyltransferase, HGPRT, HGPRTase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

13H11.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HPRT1(3251)

General description

Hypoxanthine-guanine phosphoribosyltransferase (EC:2.4.2.8; UniProt P00492; also known as HGPRT, HGPRTase) is encoded by the HPRT1 (also known as HGPRT, HPRT) gene (Gene ID 3251) in human. HPRT is a purine salvage enzyme that catalyzes the condensation of 5-phosphoribosyl 1-pyrophosphate (PRPP) and hypoxanthine or guanine to form inosine 5-monophosphate (IMP) and guanosine 5-monophosphate (GMP), respectively. PRPP is an essential compound for the de novo pathway of purine synthesis as well as the synthesis of pyrimidine compounds. PRPP concentrations in erythrocyte are markedly increased in individuals with HPRT deficiency. Complete HPRT deficiency leads to Lesch–Nyhan disease (LND) characterized by uric acid overproduction, motor dysfunction, cognitive disability, and self-injurious behavior. Partial deficiency of HPRT activity causes Lesch–Nyhan variant (LNV) characterized by hyperuricemia and variable degree of neurological manifestations, depending on the residual enzyme activity.

Immunogen

Epitope: Internal
GST-tagged human HGPRT recombinant fragment lacking N- and C-terminal regions.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-HGPRT, clone 13H11.1 is validated for use in Western Blotting for the detection of HGPRT.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected HGPRT in 10 µg of HeLa cell lysate.

Target description

~25 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service