Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2230

Sigma-Aldrich

Anti-Syntaxin 5 Antibody, clone 18C8

Synonym(s):

STX-5, Sed5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

18C8, monoclonal

species reactivity

rat

packaging

antibody small pack of 100

technique(s)

immunoprecipitation (IP): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

Gene Information

rat ... Stx5(65134)

Specificity

Clone 18C8 is a is a mouse monoclonal antibody that detects Syntaxin-5. It targets an epitope within the cytoplasmic domain in the C-terminal half. It detects both isoform 1 and 2.

Immunogen

GST-tagged recombinant fragment corresponding to 83 amino acids from the C-terminal half, SNARE motif of rat.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue extract and NRK cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Syntaxin-5 in Rat brain tissue extract and normal rat kidney epithelial (NRK) cell lysate.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated Syntaxin 5 in Immunoprecipitation applications (Williams, A.L., et al. (2004). Mol Biol Cell. 15(1):162-75).

Immunocytochemistry Analysis: A representative lot detected Syntaxin 5 in Immunocytochemistry applications (Williams, A.L., et al. (2004). Mol Biol Cell. 15(1):162-75).

Western Blotting Analysis: A representative lot detected Syntaxin 5 in Western Blotting applications (Williams, A.L., et al. (2004). Mol Biol Cell. 15(1):162-75).

Inhibition Assay: A representative lot inhibited the formation of a ternary SNARE complex. (Williams, A.L., et al. (2004). Mol Biol Cell.;15(1):162-75).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Syntaxin-5 (UniProt: Q08851; also known as STX-5) is encoded by the Stx5 (also known as Stx5a) gene (Gene ID: 65134) in rat. . Syntaxin-5 is a single-pass type IV membrane protein that localizes throughout the Golgi apparatus and is most abundant in the cis-most cisternae. It is found in the endoplasmic reticulum (ER)-Golgi intermediate compartment membrane. It has a cytoplasmic domain (aa 1-333), a transmembrane domain (aa 334-354), and a single amino acid vesicular domain (aa 355). Its expression has been reported in the brain, heart, spleen, lung, liver, kidney, and testis. It mediates ER to Golgi transport and together with vesicular transport factor p115 (USO1) and Golgi matrix protein GM130 (GOLGA2), it is involved in tethering and fusion at the cis-Golgi membrane to maintain the stacked and inter-connected structure of the Golgi apparatus. Syntaxin-5 is shown to protect cells against proteotoxicity and is involved in clearance of insoluble protein deposits. Clone 18C8 is shown to bind to syntaxin 5 SNARE motif and inhibit SNARE complex formation and prevents the SNARE motif from engaging other SNAREs. It binds to syntaxin 5 molecules with unengaged, SNARE motifs. (Ref.: Babazadeh, R., et al. (2019). Cell Rep. 28(8); 2096-2110; Diao, A., et al (2008); J. Biol. Chem. 283(11); 6957-6967; Williams, AL., et al. (2004). Mol. Biol. Cell. 15; 162-175).

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service