Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

554715-M

Millipore

Retro-2

A cell-permeable tricyclic imine compound that inhibits the endosome-to-Golgi retrograde transport, but not cellular uptake, of CtxB, StxB without affecting cellular uptake/trafficking of Tf or EGF.

Synonym(s):

Retro-2, Cholera Toxin Retrograde Transport Blocker, Ricin Retrograde Transport Blocker, SLT Retrograde Transport Blocker, 2-(((5-Methyl-2-thienyl)methylene)amino)-N-phenylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16N2OS
Molecular Weight:
320.41
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable tricyclic imine compound that inhibits the endosome-to-Golgi retrograde transport, but not cellular uptake, of CtxB (Cholera toxin B chain), StxB (Shiga-like toxin B chain), and the plant toxin ricin from castor seed in HeLa cells (20 µM), without affecting cellular uptake and trafficking of Tf (Transferrin), EGF, and vesicular stomatitis virus glycoprotein VSVGts045. Retro-2 is demonstrated to selectively affect the cellular distribution of SNARE proteins Synt5 and Synt6, but not 24 other Golgi and endocytic pathway proteins. Retro-2 pretreatment is shown to effectively protect against ricin-, Stx1-, and Stx2-induced cytotoxicity in HeLa cultures in vitro (Fold of protection against 4 h toxin incubation = 2.7, 22, and 65, respectively, with 0.5 h 20 µM Retro-2 pretreatment), as well as ricin-induced (2 µg/kg by nasal spray) fatality in mice in vivo (0 mortality with one 200 mg/kg i.p. dose 1h prior to toxin exposure).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
It is recommended to prepare a stock solution for 3 month use each time and store the unreconstituted material in solid form, protected from moisture and preferably under inert gas, for best long-term stability during storage.

Other Notes

Stechmann, B., et al. 2010. Cell141, 231.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service