Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABS2042

Sigma-Aldrich

Anti-HPx1 Antibody, clone HIC0-3B3

clone HICO-3B3, from mouse

Synonym(s):

Human pancreas exocrine cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HICO-3B3, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)

isotype

IgG1κ

NCBI accession no.

target post-translational modification

unmodified

General description

Pancreas performs both the endocrine and the exocrine function. The bulk of pancreas is composed of exocrine cells that produce digestion enzymes which are released into ducts to be emptied into the duodenum. The endocrine pancreas is composed of islets of Langerhans that produce hormones. At least five distinct endocrine cell types have been identified in the endocrine pancreas, each producing a different hormone. Beta cells produce insulin, which is considered as an anabolic hormone and regulates blood glucose levels and is also involved in fat and protein synthesis. Alpha cells produce glucagon that acts when blood glucose levels are low. Ductal cells of pancreas form the epithelial lining of the branched tubes that deliver digestive enzymes into the duodenum. They also secrete bicarbonate to neutralize acidic contents coming in from stomach. Duct cells may contribute up to 10% of all pancreatic cells. Acinar cells function as a unit of the exocrine pancreas and they synthesize, store, and secrete digestive enzymes (lipase, alpha amylase, trypsin, and chymotrypsin), which under normal physiological conditions are activated only after reaching the duodenum. Premature activation of these enzymes within pancreatic acinar cells can lead to acute pancreatitis. Clone HICO-3B3 can be used to detect exocrine cells in human pancreatic tissue. (Ref.: Dorrell, C., et al. (2008). Stem Cell Research (2008). 1(3); 183-194; Grapin-Botton, A (2005). Int. J. Biochem. Cell Biol. 37(3); 504-510; Leung, PS (2006). Int. J. Biochem. Cell Biol. 38(7); 1024-1030).

Specificity

Clone HICO-3B3 specifically detects human pancreas exocrine cells.

Immunogen

Human pancreatic enriched exocrine cells.

Application

Anti-HPx1, clone HIC0-3B3, Cat. No. MABS2042, is a mousse monoclonal antibody that detects human pancreas exocrine cells and has been tested for use in Flow Cytometry, Immunofluorescence, and Immunohistochemistry (Paraffin).
Immunofluorescence Analysis: A representative lot detected HPx1 in Immunofluorescnece applications (Dorrell, C., et. al. (2008). Stem Cell Res. 1(3):183-94).

Flow Cytometry Analysis: A representative lot detected HPx1 in Flow Cytometry applications (Dorrell, C., et. al. (2008). Stem Cell Res. 1(3):183-94; Dorrell, C., et. al. (2016). Nat Commun. 7:11756).
Research Category
Signaling

Quality

Evaluated by Immunohistochemistry (Paraffin) in human pancreatic tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected HPx1 in human pancreatic tissue sections.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service