Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABS1201

Sigma-Aldrich

Anti-OSBP Antibody, clone 1A6.2

clone 1A6.2, from mouse

Synonym(s):

Oxysterol-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A6.2, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... OSBP(5007)

Related Categories

General description

Oxysterol-binding protein 1 (UniProt P22059) is encoded by the OSBP (also known as OSBP1) gene (Gene ID 5007) in human. OSBP mediates the transport of cholesterol and phosphatidylinositol 4-phosphate (PtdIns4P, PI-4-P, PI4P) between endoplasmic reticulum (ER) and Golgi. OSBP links ER and trans-Golgi membranes at the ER-Golgi membrane contact sites (MCSs) and shuttles sterol into the Golgi and PI4P back to the ER, where it is hydrolyzed by the PI4P-phosphatase Sac1. OSBP contains an FFAT (phenylalanines (FF) in an acidic tract) motif (a.a. 358-364) that binds ER-resident VAP protein and an N-terminal pleckstrin homology (PH) domain (a.a. 88-181) that binds PI4P. OSBP and OSBP-related protein 4 (ORP4) are reported to be involved in virus-induced membrane alterations essential for viral replication organelle formation. OSBP knockdown inhibits virus replication, whereas overexpression of OSBP or ORP4 counteracts the antiviral effects of the OSBP/ORP4 antagonist OSW-1.

Specificity

Clone 1A6.2 targets an epitope in the C-terminal region of human OSBP.

Immunogen

Epitope: C-terminal region.
GST-tagged recombinant human OSBP C-terminal fragment.

Application

Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected OSBP in human cerebral cortex, small intestine, and kidney tissue sections.
Research Category
Signaling
Research Sub Category
Vesicular Trafficking
This Anti-OSBP Antibody, clone 1A6.2 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of OSBP.

Quality

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected OSBP in 10 µg of Jurkat cell lysate.

Target description

~90 kDa observed. 89.42 kDa calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service