Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MABN842

Sigma-Aldrich

Anti-PERK Antibody, clone 4F6.1

clone 4F6.1, from mouse

Synonym(s):

PERK, Eukaryotic translation initiation factor 2-alpha kinase 3, PRKR-like endoplasmic reticulum kinase, Pancreatic eIF2-alpha kinase, HsPEK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F6.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... EIF2AK3(9451)

General description

Eukaryotic translation initiation factor 2-alpha kinase 3 (EC 2.7.11.1; UniProt Q9NZJ5; also known as Eukaryotic translation initiation factor 2 alpha kinase 3, HsPEK, Pancreatic EIF2-alpha kinase, PRKR-like endoplasmic reticulum kinase) is encoded by the EIF2AK3 (also known as PEK, PERK, WRS) gene (Gene ID 9451) in human. The accumulation of unfolded or misfolded proteins in the endoplasmic reticulum (ER) induces the activation of three main unfold protein response (UPR) pathways, the inositol requiring enzyme-1 (IRE1) pathway, the activating transcription factor 6 (ATF6) pathway, and the protein kinase-like ER kinase (PERK) pathway. The PERK pathway involves PERK auto-phosphorylation/activation, resulting in PERK phosphorylation of eIF2α, which in turn leads to increased ATF4 transcription and ATF4-mediated CAAT/enhancer binding protein (C/EBP) homologous protein (CHOP) upregulation.

Immunogen

Epitope: Internal (C-terminal half)
GST-tagged recombinant protein corresponding to the Internal (C-terminal half) of human PERK.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected PERK in human liver tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-PERK Antibody, clone 4F6.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of PERK.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected PERK in 10 µg of HEK293 cell lysate.

Target description

~125 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anning Yang et al.
Journal of cellular biochemistry, 118(9), 2921-2932 (2017-02-24)
Folate deficiency is a known risk factor for liver injury; however, the underlying mechanism remains unclear. In this study, we employed a high homocysteine-induced liver injury model of Apolipoprotein E-deficient (ApoE-/- ) mice fed high-methionine diet and found that high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service