Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC955

Sigma-Aldrich

Anti-GADD153/DDIT3/CHOP Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DNA damage-inducible transcript 3 protein, DDIT-3, C/EBP zeta, C/EBP-homologous protein, CHOP, C/EBP-homologous protein 10, CHOP-10, CCAAT/enhancer-binding protein homologous protein, Growth arrest and DNA damage-inducible protein GADD153, GADD153/DDIT3/

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDIT3(1649)

General description

DNA damage-inducible transcript 3 protein (UniProt P35638; also known as CCAAT/enhancer-binding protein homologous protein, C/EBP-homologous protein 10, C/EBP zeta, CHOP, CHOP-10, DDIT-3, Growth arrest and DNA damage-inducible protein GADD153) is encoded by the DDIT3 (also known as CEBPZ, CHOP, CHOP10, GADD153) gene in human (Gene ID 1649). CHOP is a proapoptotic unfold protein response (UPR) gene and a well established marker for ER stress and UPR activation. The accumulation of unfolded or misfolded proteins in the ER induces the activation of three main UPR pathways, the inositol requiring enzyme-1 (IRE1) pathway, the activating transcription factor 6 (ATF6) pathway, and the protein kinase-like ER kinase (PERK) pathway. The PERK pathway involves PERK auto-phosphorylation/activation, resulting in PERK phosphorylation of eIF2α, which in turn leads to increased ATF4 transcription and ATF4-mediated CHOP upregulation.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to human GADD153/DDIT3/CHOP near the N-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-GADD153/DDIT3/CHOP antibody is validated for use in Western Blotting for the detection of GADD153/DDIT3/CHOP.

Quality

Evaluated by Western Blotting in Tunicamycin treated HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected GADD153/DDIT3/CHOP in 10 µg of Tunicamycin treated HeLa cell lysate.

Target description

~29 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service