Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABN624

Sigma-Aldrich

Anti-Pan-QKI Antibody, clone N147/6

clone N147/6, from mouse

Synonym(s):

Protein quaking, Hqk, HqkI, QKI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N147/6, monoclonal

species reactivity

mouse, rat, human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... QKI(9444)

General description

QKI is also called Protein Quaking or HqkI. QKI is an RNA-binding protein that plays a central role in myelinization. QKI acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation, and is itself, regulated by alternative splicing. QKI is expressed in the frontal cortex of brain, but is shown to be down-regulated in the brain of schizophrenic patients.

Specificity

This antibody demonstrates cross reactivity against QKI-6, QKI-7, and QKI-7b (Prof. J. Trimmer, University of California, Davis.).

Immunogen

Recombinant protein corresponding to human Pan-QKI.

Application

Anti-Pan-QKI Antibody, clone N147/6 is a highly specific mouse monoclonal antibody & that targets QKI & has been tested in western blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Pan-QKI in human frontal cortex.

Immunohistochemistry Analysis: A representative lot detected Pan-QKI in rat hippocampus tissue (Prof. J. Trimmer, University of California, Davis.).

Western Blotting Analysis: 2 µg/mL from a representative lot detected Pan-QKI in 10 µg of rat brain tissue.

Western Blotting Analysis: 2 µg/mL from a representative lot detected Pan-QKI in 10 µg of mouse brain membrane tissue.

Western Blotting Analysis: A representative lot detected Pan-QKI in RBM cell lysates and in COS cell lysates transfected with QKI-5, QKI-6, QKI-7, and QKI-7b (Prof. J. Trimmer, University of Califorina, Davis.).
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in rat brain membrane tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Pan-QKI in 10 µg of rat brain membrane tissue lysate.

Target description

~36-40 kDa observed. Uniprot describes six isoforms observed between 35-38 kDa Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain membrane tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Best et al.
Nature communications, 15(1), 6328-6328 (2024-07-28)
Disruption of alternative splicing frequently causes or contributes to human diseases and disorders. Consequently, there is a need for efficient and sensitive reporter assays capable of screening chemical libraries for compounds with efficacy in modulating important splicing events. Here, we
Hiroaki Hojo et al.
The Journal of biological chemistry, 295(2), 390-402 (2019-12-04)
MicroRNA-122 (miR-122) is highly expressed in hepatocytes, where it plays an important role in regulating cholesterol and fatty acid metabolism, and it is also a host factor required for hepatitis C virus replication. miR-122 is selectively stabilized by 3' adenylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service