Skip to Content

AB9904

Anti-QKI-5 Antibody

serum, Chemicon®

Synonym(s):

Quaking Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
13
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... QKI(9444)

General description

QKI is an RNA-binding protein that plays a central role in myelinization. Acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation. It is regulated by alternative splicing.
~40 kDa

Immunogen

Synthetic peptide from mouse QKI-5.

Application

Detect QKI-5 using this Anti-QKI-5 Antibody validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

RNA Binding Protein (RBP)

Biochem/physiol Actions

QKI-5 (Quaking Protein). The antibody recognizes the ~40 kDa protein.

Physical form

Liquid.
Serum

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Positive Control: Glial cells, astrocytes and oligodendrocytes.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library