Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN291

Sigma-Aldrich

Anti-IMPDH1 Antibody, clone 4F9.1

clone 4F9.1, from mouse

Synonym(s):

Inosine-5′-monophosphate dehydrogenase 1, IMP dehydrogenase 1, IMPD 1, IMPDH 1, IMPDH-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F9.1, monoclonal

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IMPDH1(3614)

General description

Inosine-5′-monophosphate dehydrogenase 1 (IMPDH 1) catalyzes the conversion of inosine 5′-phosphate (IMP) to xanthosine 5′-phosphate (XMP), as part of the de novo synthesis of guanine nucleotides. It therefore is crucial in the regulation of cell growth. IMPDH 1 may also have single-stranded nucleic acid-binding activity involving it in RNA and/or DNA metabolism, and is suspected to play a role in the growth progression of some tumors. Defects in IMPDH 1 abnormalities are linked to retinitis pigmentosa type 10 (RP10) and Leber congenital amaurosis type 11 (LCA11). Due to the connection between IMPDH activity and cell proliferation, it has been become a target for cancer and viral chemotherapy and immunosuppressive drugs. The activities of the antitumor drug tiazofurin, the antiviral drug ribavirin, and the immunosuppressive drugs mizoribine and mycophenolic acid (MPA) are attributed to the inhibition of IMPDH. In addition, bacterial and parasitic IMPDH′s differ significantly from mammalian enzymes, which make it a suitable target for anti-infective drugs.

Immunogen

GST-tagged recombinant protein corresponding to human IMPDH1.

Application

Anti-IMPDH1 Antibody, clone 4F9.1 is a highly specific mouse monoclonal antibody, that targets Inosine-5′-monophosphate dehydrogenase 1 & has been tested in western blotting & IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected IMPDH1 in 10 µg of NIH3T3 cell lysate.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected IMPDH1 in 10 µg of Jurkat cell lysate.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected IMPDH1 in rat eye tissue.

Quality

Evaluated by Western Blotting in PC-12 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected IMPDH1 in 10 µg of PC-12 cell lysate.

Target description

~55 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC-12 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service