Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

ZRB2544

Sigma-Aldrich

Anti-MAVS Antibody, clone 1L3 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CARD adapter inducing interferon beta, IPS1, Interferon beta promoter stimulator protein 1 (IPS-1), KIAA1271, Putative NF-kappa-B-activating protein 031N, Virus-induced-signaling adapter (VISA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1L3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 56.5KDa kDa
observed mol wt ~62 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

N-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... MAVS(57506)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1L3 is a ZooMAb® rabbit recombinant monoclonal antibody that detects MAVS. It targets an epitope within 23 amino acids from the N-terminal cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the N-terminal cytoplasmic domain of human MAVS.

Application

Quality Control Testing

Evaluated by Western Blotting in U-2 OS cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected MAVS in U-2 OS cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound MAVS peptide with at least one thousand-fold (1,000X)) higher than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected MAVS in human kidney tissue sections.

Immunocytochemistry Analysis: Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected MAVS in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Mitochondrial antiviral-signaling protein (UniProt: Q7Z434; also known as CARD adapter inducing interferon beta;Interferon beta promoter stimulator protein 1 (IPS-1), Putative NF-kappa-B-activating protein 031N, Virus-induced-signaling adapter (VISA)) is encoded by the MAVS (also known as IPS1, KIAA1271, VISA) gene (Gene ID: 57506) in human. Mitochondrial antiviral-signaling protein (MAVS) is a key adaptor molecule in the innate immune response to viral infection. It is characterized by a caspase activation and recruitment domain (CARD) at its N-terminus, a proline-rich region, and a C-terminal transmembrane domain that anchors it to the outer mitochondrial membrane. The CARD domain is crucial for its interaction with other CARD-containing proteins like RIG-I and MDA5, which detect viral RNA. MAVS plays a role the signal transduction from RIG-I-like receptors (RLRs) that recognize viral RNA, leading to the activation of transcription factors such as NF- B and IRFs (Interferon Regulatory Factors). This results in the expression of type I interferons and other inflammatory cytokines, which are essential components of the antiviral response. MAVS is expressed in a variety of cell types, reflecting its broad role in the innate immune response. MAVS has been implicated in several diseases due to its central role in antiviral immunity. Inefficient MAVS signaling can lead to increased susceptibility to viral infections, while overactive MAVS signaling can result in excessive inflammation. MAVS activity has been associated with autoimmune diseases (type I diabetes, systemic lupus erythematosus) and chronic inflammatory diseases such as hepatitis.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service