Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABN2728

Sigma-Aldrich

Anti-FoxA2/HNF-3b Antibody, clone 4C7

Synonym(s):

Forkhead box A2, HNF-3 beta, FOXA2, Hepatocyte nuclear factor-3beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4C7, monoclonal

mol wt

calculated mol wt 47.23 kDa
observed mol wt ~50 kDa

purified by

using protein G

species reactivity

human, chicken, mouse

packaging

antibody small pack of 100

technique(s)

electron microscopy: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

chicken ... HNF-3beta(395539)

Specificity

Clone 4C7 is a mouse monoclonal antibody that detects Hepatocyte nuclear factor-3β (HNF-3β/FoxA2).

Immunogen

Recombinant Chicken Rhodopsin (1-6).

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected FoxA2/HNF-3b, in HepG2 cell lysate.

Tested Applications

Electrophoretic Mobility Shift Assay: A representative lot detected FoxA2/HNF-3b in Electrophoretic Mobility Shift Assay (EMSA) (Foucher, I., et al. (2003). Development. 130(9): 1867-76).

Immunofluorescence Analysis: A representative lot detected FoxA2/HNF-3b in Immunofluorescence applications (Camp, D., et al. (2014). Development. 141(20): 3879-88; Hoelzl, M.A., et al. (2017). Dev Biol. 429(1): 132-146).

Immunoprecipitation Analysis: A representative lot immunoprecipitated FoxA2/HNF-3b in Immunoprecipitation applications (van den Brink, G.R., et al. (2001). Gastroenterology. 121(2):317-28).

Immunohistochemistry Applications: A representative lot detected FoxA2/HNF-3b in Immunohistochemistry applications (Ericson, J., et al. (1996). Cell. 87(4):661-73; Kwon, O., et al. (1998). J Clin Invest. 101(10):2054-64; Rousa, F.M., et al. (2000). Mol Cell Biol. 20(21):8264-82; van den Brink, G.R., et al. (2001). Gastroenterology. 121(2):317-28; Camp, D., et al. (2014). Development. 141(20):3879-88; Hoelzl, M.A., et al. (2017). Dev Biol. 429(1):132-146; Amarnath, S., et al. (2017). J Cell Sci. 130(1):119-131).

Immunocytochemistry Analysis: A representative lot detected FoxA2/HNF-3b in Immunohistochemistry applications (Nagase, T., et al. (2009). Dev Dyn. 238(5):1118-30).

Western Blotting Analysis: A representative lot detected FoxA2/HNF-3b in Western Blotting applications (Rousa, F.M., et al. (2000). Mol Cell Biol. 20(21):8264-82; van den Brink, G.R., et al. (2001). Gastroenterology. 121(2):317-28).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Hepatocyte nuclear factor-3beta (UniProt: Q9PWP8; also known as HNF-3 beta, FOXA2) is encoded by the HNF-3beta gene (Gene ID: 395539) in Chicken. FoxA2 belongs to the forkhead family of transcription factors that uses a modified helix-turn-helix motif to bind DNA as a monomer. Its DNA binding domain is localized in amino acids 151-245. It is expressed very early during development and is involved in the development of several tissues, including liver, pancreas, lung, prostate, and the nervous system. In mouse embryo, FoxA2 expression is initiated during gastrulation in the node, notochord, and floor plate. Its induction in neural plate cells is an early response to notochord-derived signals and its ectopic expression induces floor plate differentiation. In the ventral mesencephalon and cerebellum, it is co-expressed with Engrailed homeoproteins - En1 and En2, which raises the possibility that the two transcription factors interact to regulate common target genes. FoxA2 expression is observed in different regions of the developing nervous system. Along with FoxA1, it is reported to be crucial for the specification and differentiation of dopamine neurons during embryonic development. FoxA2 also functions as a suppressor of tumor metastasis by inhibition of epithelial-to-mesenchymal transition in human lung cancers. It is shown to be a key target of TGF-β1 in controlling epithelial-to-mesenchymal transition (EMT) in lung cancer cells. (Ref.: Rausa, FM., et al. (2020). Mol. Cell. Biol. 20(21); 8264-8282; Domanskyi, A., et al (2014). Front. Cell. Neurosci. 8; 275; Tang, Y., et al. (2011). Cell Res. 21(2); 316-326; Foucher, I., et al. (2003). Development 130(9); 1867-1876; Ericson, J., et al. (1996). Cell. 87(4); 661-673).

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service