Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

ZRB1872

Sigma-Aldrich

Anti-FOXA2 Antibody, clone 1H12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Hepatocyte nuclear factor 3-beta;HNF-3-beta;HNF-3B;Forkhead box protein A2;Transcription factor 3B;TCF-3B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H12, recombinant monoclonal

description

1H12 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 48.31 kDa
observed mol wt ~52 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

rat, canine, feline, monkey, bovine, porcine, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FOXA2(3170)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects FOXA2. It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human FOXA2.

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected FOXA2 in A549 cell lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected FOXA2 in A549 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FOXA2 in human colon tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected FOXA2 in A549 cell lysate.

Target description

Hepatocyte nuclear factor 3-beta (UniProt: Q9Y261; also known as HNF-3-beta, HNF-3B, Fork head box protein A2, Transcription factor 3B, TCF-3B) is encoded by the FOXA2 (also known as HNF3B, TCF3B) gene (Gene ID: 3170) in human. The FOX family of transcription factors contains more than 50 members, all of which share a winged-helix DNA binding domain. FOXA2 is a fork-head box transcription factor that is involved in embryonic development, establishment of tissue-specific gene expression, and regulation of gene expression in differentiated tissues. It is required for notochord formation and is involved in the development of multiple endoderm-derived organ systems such as the liver, pancreas, and lungs. It contains two transactivation domains (aa 14-93 and 361-457) and its fork-head domain is localized in amino acids 159-252. It shuttles between the nucleus and cytoplasm in a CRM1-dependent manner and in response to insulin signaling via AKT1 it is exported from the nucleus. Phosphorylation of FOXA2 at threonine 156 by AKT inactivates its transcription function and induces its nuclear exclusion. Deletion of FOXA2 in the epithelial cells of the developing mouse lung is shown to inhibit the differentiation of respiratory epithelial cells and causes neonatal respiratory failure, with the characteristics of respiratory distress syndrome in preterm infants. FOXA2 knockdown is reported to induce EMT and increase invasion of A549 lung cancer cells and overexpression of FOXA2 inhibits human lung cancer cell invasion and migration. Two isoforms of FOXA2 have been described that are produced by alternative splicing. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Li, J., et al. (2017). Biochemistry. 56(29); 3745-3753; Tang, Y., et al. (2011). Cell Res. 21; 316-326).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service