Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABN2394

Sigma-Aldrich

Anti-Syntaxin-3B Antibody, clone 12E5

clone 12E5, from mouse

Synonym(s):

STX3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

12E5, monoclonal

species reactivity

mouse, human, bovine

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Stx3(20908)

General description

Syntaxin-3 (UniProt: Q64704; also known as STX3) is encoded by the Stx3 gene (Gene ID: 20908) in murine species. Syntaxin-3B is a single-pass type IV membrane protein that is specific for ribbon synapses of the retina. Stx3 gene codes for four different splice forms, Syntaxin 3A, 3B, 3C and 3D. Syntaxin 3A and 3B have an identical N-terminal domain but differ in the C-terminal half of the SNARE domain and the C-terminal transmembrane domain. These two domains are thought to be directly involved in synaptic vesicle fusion. Syntaxin 3 protein is found in all ribbon synapses of the retina. This type of synapse in the retina is formed by rod and cone photoreceptors, as well as by bipolar neurons. Syntaxin 3B has been shown to be important for synaptic vesicle exocytosis in retinal bipolar cells indicating a specific role for Syntaxin 3B in ribbon synapses. Syntaxin 3B can be phosphorylation at Thr14 by CAM Kinase II is shown to increase its binding to SNAP-25. (Ref.: Curtis, LB et al. (2008). J. Comp. Neurol. 510(5): 550-559; Liu, X et al. (2014). Mol. Cell. Neurosci. 60(1): 53-62).

Specificity

Clone 12E5 is a highly specific antibody against the B isoform of Syntaxin 3, which is abundantly expressed in photoreceptors of the retina.

Immunogen

GST-tagged recombinant murine Syntaxin 3, isoform B that lacks the transmembrane domain.

Application

Anti-Syntaxin-3B clone 12E5, Cat. No. MABN2394, is a highly specific mouse monoclonal antibody that targets Syntaxin 3B and has been tested in Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Syntaxin-3B in human retina and mouse eye tissues.

Western Blotting Analysis: A representative lot detected Syntaxin-3B in Western Blotting applications (Zulliger, R., et. al. (2015). PLoS One. 10(9):e0138508).

Immunoprecipitation Analysis: A representative lot detected Syntaxin-3B in Immunoprecipitation applications (Zulliger, R., et. al. (2015). PLoS One. 10(9):e0138508).

Immunohistochemistry Analysis: A representative lot detected Syntaxin-3B in Immunohistochemistry applications (Zulliger, R., et. al. (2015). PLoS One. 10(9):e0138508).

Immunocytochemistry Analysis: A representative lot detected Syntaxin-3B in Immunocytochemistry applications (Zulliger, R., et. al. (2015). PLoS One. 10(9):e0138508).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human retinal tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Syntaxin-3B in 10 µg of human retinal tissue lysate.

Target description

~33 kDa observed; 32.95 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service