Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABN2308

Sigma-Aldrich

Anti-TrkA Antibody, clone MNAC13

Synonym(s):

High affinity nerve growth factor receptor;EC:2.7.10.1;Neurotrophic tyrosine kinase receptor type 1;TRK1-transforming tyrosine kinase protein;Tropomyosin-related kinase A;Tyrosine kinase receptor;Tyrosine kinase receptor A;gp140trk;p140-TrkA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MNAC13, monoclonal

mol wt

calculated mol wt 87.5 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

ELISA: suitable
neutralization: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NTRK1(4914)

General description

High affinity nerve growth factor receptor (UniProt: P04629; also known as EC:2.7.10.1, Neurotrophic tyrosine kinase receptor type 1, TRK1-transforming tyrosine kinase protein, Tropomyosin-related kinase A, Tyrosine kinase receptor, Tyrosine kinase receptor A, Trk-A, gp140trk, p140-TrkA) is encoded by the NTRK1 (also known as MTC, TRK, TRKA) gene (Gene ID: 4914) in human. Trk-A is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-32), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 33-423), a transmembrane domain (aa 424-439), and a cytoplasmic domain (aa 440-796). The extracellular domain of Trk-A is composed by five domains: a leucine-rich region, two cysteine-rich domains and two immunoglobulin (Ig)-like domains. Trk-A serves as a receptor tyrosine kinase involved in the development and the maturation of the central and peripheral nervous system through regulation of proliferation, differentiation, and survival of sympathetic and nervous neurons. It serves as a high affinity receptor for NGF, which is its primary ligand. It can also bind and be activated by neurotrophin-3 (NTF3). However, NTF3 only supports axonal extension through NTRK1 but does not affect neuron survival. Upon dimeric NGF ligand-binding, it undergoes homodimerization, autophosphorylation. and activation. Clone MNAC13 recognizes Trk-A with high affinity and specificity and displays a neutralizing activity toward the NGF/TrkA interaction. (Ref.: Covaceuszach, S., et al. (2005). Proteins. 58(3); :717-727; Jing, S., et al. (1992). Neuron. 9(6); 1067-1079).

Specificity

Clone MNAC13 is a mouse monoclonal antibody that detects TrkA. It targets an epitope within 17 amino acids within the first Ig-like C2 type domain from the extracellular domain.

Immunogen

BALB/C 3T3-transfected cells expressing human Trk-A protein.

Application

Quality Control Testing

Evaluated by ELISA with recombinant Human TrkA.

ELISA Analysis (ELISA): Various dilutions of this antibody detected recombinant Human TrkA.

Tested applications

ELISA Analysis: A representative lot detected TrkA in ELISA applications (Covaceuszach, S., et al. (2005). Proteins. 58(3):717-27).

Neutralizing: A representative lot neutralized TrkA activity in Neutralizing applications (Covaceuszach, S., et al. (2005). Proteins. 58(3):717-27; Ugolini, G., et al. (2007). Proc Natl Acad Sci USA. 104(8):2985-90).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TrkA, clone MNAC13, Cat. No. MABN2308, is a mouse monoclonal antibody that detects TrkA and is tested for use in ELISA and Neutralizing.

Physical form

Purified mouse monoclonal antibody IgG1 in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service