Skip to Content
MilliporeSigma
All Photos(12)

Documents

ZRB1639

Sigma-Aldrich

Anti-TrkA Antibody, clone 2J4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

High affinity nerve growth factor receptor;EC:2.7.10.1;Neurotrophic tyrosine kinase receptor type 1;TRK1-transforming tyrosine kinase protein;Tropomyosin-related kinase A;Tyrosine kinase receptor;Tyrosine kinase receptor A;gp140trk;p140-TrkA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2J4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 87.5 kDa
observed mol wt ~140 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... NTRK1(4914)

General description

ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2J4 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects TrkA . It targets an epitope within 21 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal, cytoplasmic domain of human TrkA.

Application

Quality Control Testing

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected TrkA in K562 cell lysate.

Tested Applications

Western Blotting Analysis (WB): 1:10,000 dilution of this antibody detected His-tagged recombinant fragment of Human TrkA.

Affinity Binding Assay: A representative lot of this antibody bound TrkA peptide with a KD of 4.3 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TrkA in SH-SY5Y cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TrkA in human adrenal gland tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

High affinity nerve growth factor receptor (UniProt: P04629; also known as EC:2.7.10.1, Neurotrophic tyrosine kinase receptor type 1, TRK1-transforming tyrosine kinase protein, Tropomyosin-related kinase A, Tyrosine kinase receptor, Tyrosine kinase receptor A, Trk-A, gp140trk, p140-TrkA) is encoded by the NTRK1 (also known as MTC, TRK, TRKA) gene (Gene ID: 4914) in human. TrkA is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-32), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 33-423), a transmembrane domain (aa 424-439), and a cytoplasmic domain (aa 440-796). The extracellular domain of Trk-A is composed by five domains: a leucine-rich region, two cysteine-rich domains and two immunoglobulin (Ig)-like domains. TrkA serves as a receptor tyrosine kinase involved in the development and the maturation of the central and peripheral nervous system through regulation of proliferation, differentiation, and survival of sympathetic and nervous neurons. It serves as a high affinity receptor for NGF, which is its primary ligand. It can also bind and be activated by neurotrophin-3 (NTF3). However, NTF3 only supports axonal extension through NTRK1 but does not affect neuron survival. Upon dimeric NGF ligand-binding, it undergoes homodimerization, autophosphorylation. and activation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Wehrman, T., et al. (2007). Neuron. 53(1); 25-38; Covaceuszach, S., et al. (2005). Proteins. 58(3); :717-727; Jing, S., et al. (1992). Neuron. 9(6); 1067-1079).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service