Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

MABN2297

Sigma-Aldrich

Anti-Amyloid Precursor Protein (APP) Antibody

rat monoclonal, 7H6

Synonym(s):

Amyloid beta A4 protein, ABPP, APPI, Alzheimer disease amyloid protein, Amyloid precursor protein, Beta-amyloid precursor protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-APP Antibody, clone 7H6, clone 7H6, from rat

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7H6, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Related Categories

General description

Amyloid beta A4 protein (UniProt: P05067; also known as ABPP, APPI, APP, Alzheimer disease amyloid protein, Amyloid precursor protein, Beta-amyloid precursor protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II) is encoded by the APP (also known as A4, AD1) gene (Gene ID: 351) in human. APP undergoes extensive post-translational modification including glycosylation, phosphorylation, and tyrosine sulfation, as well as many types of proteolytic processing to generate peptide fragments. APP is proteolytically processed under normal cellular conditions by alpha-secretase or beta-secretase to generate and release soluble APP peptides, S-APP-alpha and S-APP-beta, and the retention of corresponding membrane-anchored C-terminal fragments, C80, C83 and C99. Subsequent processing of C80 and C83 by gamma-secretase yields P3 peptides. In Alzheimer s disease processing of C99 generates amyloid-beta 40 (Abeta40) and amyloid-beta 42 (Abeta42) that form amyloid plaques. Beta-amyloid peptides are lipophilic metal chelators with metal-reducing activity. They bind transient metals such as copper, zinc and iron. APP can also be cleaved by caspases during neuronal apoptosis. Cleavage at Asp-739 by either caspase-6, -8 or -9 results in the production of the neurotoxic C31 peptide and the increased production of beta-amyloid peptides. In addition to its obvious role in Alzheimer′s disease, the most-substantiated role for APP is in synaptic formation and repair. Its expression is upregulated during neuronal differentiation and after neural injury.

Specificity

Clone 7H6 specifically detects human amyloid precursor protein (APP) and does not display reactivity with other species. It targets an epitope within the ectodomain region of APP.

Immunogen

StrepII-tagged human recombinant ectodomain region of the neuronal isoform hAPP695 lacking the KPI domain.

Application

Anti-APP, clone 7H6, Cat. No. MABN2297, is a highly specific rat monoclonal antibody, that targets Amyloid beta A4 protein and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected APP in HEK293 cells.

Western Blotting Analysis: A representative lot detected APP in Western Blotting applications (Hoefling, C., et. al. (2016). Aging Cell. 15(5):953-63).

Flow Cytometry Analysis: A representative lot detected APP in Flow Cytometry applications (Hoefling, C., et. al. (2016). Aging Cell. 15(5):953-63).

Western Blotting Analysis: A 1:10 dilution from a representative lot detected App in WT and APP knockdown HEK293T cells (Courtesy of Dr. med. Peer-Hendrik Kuhn, Ph.D., Institut fur Allgemeine Pathologie und Pathologische Anatomie, Technische Universität München, Munich, Germany).

Immunoprecipitation Analysis: A representative lot detected APP in Immunoprecipitation applications (Hoefling, C., et. al. (2016). Aging Cell. 15(5):953-63).

ELISA Analysis: A representative lot detected APP in ELISA applications (Hoefling, C., et. al. (2016). Aging Cell. 15(5):953-63).

Immunocytochemistry Analysis: A 1:10 dilution from a representative lot detected APP in HEK293T either lentivirally transduced with APP shRNA-1 or APP shRNA-2 both coexpressing GFP as a reporter (Courtesy of Dr. med. Peer-Hendrik Kuhn, Ph.D., Institut fur Allgemeine Pathologie und Pathologische Anatomie, Technische Universität München, Munich, Germany).

Immunocytochemistry Analysis: A representative lot detected APP in Immunocytochemistry applications (Hoefling, C., et. al. (2016). Aging Cell. 15(5):953-63).
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected APP in 10 µg of HEK293 cell lysate.

Target description

~95 kDa observed; 86.94 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service