Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS2207

Sigma-Aldrich

Anti-Inhibin α chain (INHA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.74

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody
serum

antibody product type

primary antibodies

clone

polyclonal

mol wt

calculated mol wt 39.19 kDa

purified by

(Unpurified)

species reactivity

porcine, mouse, rat

packaging

antibody small pack of 100 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Porcine ... INHA(397386)

Related Categories

General description

Inhibin alpha chain (UniProt: P04087; also known as IHNA, A-inhibin subunit) is encoded by the INHA gene (Gene ID: 397386) in porcine. Inhibins are dimeric glycoproteins that are composed of an -subunit and either a βA (inhibin A) or a βB-subunit (inhibin B). Inhibins and activins inhibit and activate, respectively, the secretion of follicle stimulating hormone (FSH) by the pituitary gland. They are also involved in regulating a number of other functions. Inhibins are glycoprotein hormones of the TGFβ superfamily and are composed of two subunits, an -subunit (20 kDa) and a β-subunit (13 kDa), linked by a disulfide bridge. Two forms of inhibin have been identified, Inhibin-A and -B. Their subunits are shown to be identical, while the β-subunit differs between both forms of inhibin and are denoted as βA and βB, respectively. Inhibin-A is secreted during the luteal phase, while Inhibin-B is predominantly secreted during the follicular phase. The inhibin -subunit contains an N-linked glycosylation site and this glycosylation is shown to be essential for its FSH suppressing activity. Porcine Inhibin chain is synthesized as a protein of 364 amino acids with a signal peptide (aa 1-17) and a propeptide (aa 18-60), which are subsequently cleaved off to generate Inhibin N-terminal region (aa 61-230) and the Inhibin chain (aa 231-364). Subsequent cleavage at aa 230-231 results in the production of Inhibin chain. Circulating level of Inhibins is a reliable marker for some types of ovarian cancer and is a predictor for the conditions of spermatogenesis. (Ref.: Makanji, Y., et al. (2014). Endocr. Rev. 35(5); 747-794; Bilezikjian, LM., et al. (1993). Endocrinology. 133(6); 2545-2553).

Specificity

This rabbit polyclonal antibody detects Inhibin chain. It targets an epitope within the first 26 amino acids from the mature Inhibin chain.

Immunogen

Linear peptide corresponding to the first 26 amino acids from the mature Inhibin chain of Porcine (aa 231-256).

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in porcine testis tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected Inhibin chain (INHA) in porcine testis tissue sections.

Tested Applications

Immunoprecipitation Analysis: A representative lot immunoprecipitated Inhibin chain (INHA) in Immunoprecipitation applications (Bilezikijan, L.M., et. al. (1993). Endocrinology. 133(6):2545-53).

Immunohistochemistry Applications: A representative lot detected Inhibin chain (INHA) in Immunohistochemistry applications (Kim, O., et. al. (2020). Proc. Natl. Acad. Sci. USA. 117(50);:31993-32004).

Western Blotting Analysis: A representative lot detected Inhibin chain (INHA) in Western Blotting applications (Bilezikijan, L.M., et. al. (1993). Endocrinology. 133(6):2545-53).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Inhibin chain (INHA), Cat. No. ABS2207, is a rabbit polyclonal antibody that detects Inhibin a chain and is tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service