Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE284

Sigma-Aldrich

Anti-MSH2 Antibody, clone FE11

clone FE11, from mouse

Synonym(s):

DNA mismatch repair protein Msh2, hMSH2, MutS protein homolog 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FE11, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MSH2(4436)

General description

MSH2 is a ubiquitously expressed nuclear protein that is involved in DNA repair processes. MSH2 is active as a heterodimer of MutS alpha and beta subunits that bind DNA at mismatched strands. The MutS alpha subunit binds to single base mismatches and dinucleotide insertion-deletion loops, whereas MutS beta detects longer insertion-deletion loops. Bound MutS subunits then form complexes with MutL alpha dimers, to coordinate downstream DNA-mismatch–repair processes. Abnormal expression of MSH2 has been linked to hereditary non-polyposis colorectal cancer type 1 and Muir-Torre syndrome.

Specificity

This antibody recognizes the C-terminus of MSH2.

Immunogen

Epitope: C-terminus
Recombinant protein corresponding to the C-terminus of human MSH2.

Application

Anti-MSH2 Antibody, clone FE11 is a Mouse Monoclonal Antibody for detection of MSH2 also known as DNA mismatch repair protein Msh2, hMSH2, MutS protein homolog 2 & has been validated in WB & IHC.
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected SW-480 in human normal colonic tissue and in human adenocarcinoma tissue (Thibodeau, S. N., et al. (1996). 56(21):4836-4840.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blot in SW-480 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected MSH2 in 10 µg of SW-480 cell lysate.

Target description

~104 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
SW-480 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hauke Schoop et al.
Translational oncology, 13(2), 165-176 (2019-12-23)
With recent studies uncovering the complex landscape of immune checkpoint regulators in gastric cancer (GC), we aimed to characterize the expression of the checkpoint proteins V-domain Ig suppressor of T-cell activation (VISTA), programmed cell death 1 ligand 1 (PD-L1), and
Jonathan A Nowak et al.
The Journal of molecular diagnostics : JMD, 19(1), 84-91 (2016-11-20)
Mismatch repair protein deficiency (MMR-D) and high microsatellite instability (MSI-H) are features of Lynch syndrome-associated colorectal carcinomas and have implications in clinical management. We evaluate the ability of a targeted next-generation sequencing panel to detect MMR-D and MSI-H based on mutational
Ester Borras et al.
Cancer prevention research (Philadelphia, Pa.), 10(10), 580-587 (2017-08-03)
Lynch syndrome (LS) is a genetic condition secondary to germline alterations in the DNA mismatch repair (MMR) genes with 30% of changes being variants of uncertain significance (VUS). Our aim was to perform an in silico reclassification of VUS from
V S Warneke et al.
British journal of cancer, 109(8), 2217-2227 (2013-09-07)
We investigated the expression of members of the epithelial cell adhesion molecule (EpCAM) signalling pathway in gastric cancer (GC) testing the following hypotheses: are these molecules expressed in GC and are they putatively involved in GC biology. The study cohort
Mev Dominguez-Valentin et al.
BMC urology, 16, 15-15 (2016-03-26)
A possible role for prostate cancer in Lynch syndrome has been debated based on observations of mismatch-repair defective tumors and reports of an increased risk of prostate cancer in mutation carriers. Potential inclusion of prostate cancer in the Lynch syndrome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service