Skip to Content
MilliporeSigma
All Photos(2)

Documents

MABE1103

Sigma-Aldrich

Anti-CST complex subunit CTC1 Antibody, clone C482

clone C482, from mouse

Synonym(s):

CST complex subunit CTC1, Conserved telomere maintenance component 1, HBV DNAPTP1-transactivated protein B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C482, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTC1(80169)

General description

CST complex subunit CTC1 (UniProt Q2NKJ3; also known as Conserved telomere maintenance component 1, HBV DNAPTP1-transactivated protein B) is encoded by the CTC1 (also known as C17orf68, CRMCC) gene (Gene ID 80169) in human. CTC1 is a subunit of the trimeric CST (CTC1, STN1, and TEN1) complex that binds ssDNA, promotes telomere DNA synthesis, and inhibits telomerase-mediated telomere elongation. CST directly interacts with and stimulates DNA polymerase -primase (pol -primase) activity. CST also associates with and sequesters telomeric 3′ overhangs after their elongation by telomerase, thereby limiting further telomere extension by telomerase. Knockdown of either STN1 or CCT1 in human cancer cells and in mouse fibroblasts results in substantially elongated telomeric G-ove+D13rhangs. Conditional deletion of CTC1 in mice leads to early entry into senescence and signs of telomeric replication defects, Human CTC1 gene mutations casue dyskeratosis congenita (DC) and related bone marrow failure syndromes, as well as the rare autosomal recessive neurological disorder Coats Plus, where patients develop cranial calcifications and cysts. Coats Plus and DC patients develop premature hair graying, anemia, and osteoporosis as a result of telomerase defects.

Specificity

Clone C482 targets an N-terminal region sequence present in both human CTC1 spliced isoforms reported by UniProt (Q2NKJ3).

Immunogen

Linear peptide corresponding to an N-terminal region sequence of human CTC1.

Application

Anti-CST complex subunit CTC1, clone C482, Cat. No. MABE1103, is a highly specific mouse monoclonal antibody that targets CTC1 and has been tested in and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: 0.75-1 µg/mL from a representative lot detected the exogenously overexpressed CTC1 in 100-150 ng of lysate from transfected HeLa cells, as well as downregulated CTC1 expression following conditional CTC1 gene knockout in 150 ng of lysate from a HCT116 cell line harboring floxed CTC1 alleles (Courtesy of Carolyn Price, Ph.D. and Xuyang Feng, Ph.D., University of Cincinnati College of Medicine, U.S.A.).

Quality

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be IgG2b .

Target description

~135 kDa observed. 134.61/114.1 kDa (isoform 1/2) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuyang Feng et al.
Nucleic acids research, 45(8), 4281-4293 (2017-03-24)
To prevent progressive telomere shortening as a result of conventional DNA replication, new telomeric DNA must be added onto the chromosome end. The de novo DNA synthesis involves elongation of the G-rich strand of the telomere by telomerase. In human
Zachary Mirman et al.
Nature, 560(7716), 112-116 (2018-07-20)
In DNA repair, the resection of double-strand breaks dictates the choice between homology-directed repair-which requires a 3' overhang-and classical non-homologous end joining, which can join unresected ends1,2. BRCA1-mutant cancers show minimal resection of double-strand breaks, which renders them deficient in
Xuyang Feng et al.
Nature communications, 9(1), 2827-2827 (2018-07-22)
Telomerase elongates the telomeric G-strand to prevent telomere shortening through conventional DNA replication. However, synthesis of the complementary C-strand by DNA polymerase α is also required to maintain telomere length. Polymerase α cannot perform this role without the ssDNA binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service