Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABE1015

Sigma-Aldrich

Anti-CST complex subunit CTC1 Antibody, clone 8E5.2

clone 8E5.2, from mouse

Synonym(s):

CST complex subunit CTC1, Conserved telomere maintenance component 1, HBV DNAPTP1-transactivated protein B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8E5.2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CTC1(80169)

General description

CST complex subunit CTC1 (UniProt Q2NKJ3; also known as Conserved telomere maintenance component 1, HBV DNAPTP1-transactivated protein B) is encoded by the CTC1 (also known as C17orf68, CRMCC) gene (Gene ID 80169) in human. CTC1 is a subunit of the trimeric CST (CTC1, STN1, and TEN1) complex that binds ssDNA, promotes telomere DNA synthesis, and inhibits telomerase-mediated telomere elongation. CST directly interacts with and stimulates DNA polymerase α-primase (polα-primase) activity. CST also associates with and sequesters telomeric 3′ overhangs after their elongation by telomerase, thereby limiting further telomere extension by telomerase. Knockdown of either STN1 or CCT1 in human cancer cells and in mouse fibroblasts results in substantially elongated telomeric G-overhangs. Conditional deletion of CTC1 in mice leads to early entry into senescence and signs of telomeric replication defects, Human CTC1 gene mutations casue dyskeratosis congenita (DC) and related bone marrow failure syndromes, as well as the rare autosomal recessive neurological disorder Coats Plus, where patients develop cranial calcifications and cysts. Coats Plus and DC patients develop premature hair graying, anemia, and osteoporosis as a result of telomerase defects.

Specificity

Expected to react with both spliced isoforms of human CTC1 reported by UniProt (Q2NKJ3).

Immunogen

Epitope: Internal (C-terminal half).
GST-tagged recombinant internal fragment from the C-terminal half of human CTC1.

Application

Anti-CST complex subunit CTC1 Antibody, clone 8E5.2 is an antibody against CST complex subunit CTC1 for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Western Blotting in human cortex tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected CST complex subunit CTC1 in 10 µg of human cortex tissue lysate.

Target description

~135 kDa observed. 134.6 kDa (isoform 1) and 114.1 kDa (isoform 2) calculated. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service