Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE1081

Sigma-Aldrich

Anti-5-Methylcytosine (5mC) Antibody, clone EDL MC-4

clone EDL MC-4, from mouse

Synonym(s):

5mC, 5-methylcytosine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EDL MC-4, monoclonal

species reactivity (predicted by homology)

all

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable

isotype

IgG2aλ

shipped in

ambient

target post-translational modification

unmodified

General description

Four types of cytosine modifications have been identified in mammalian DNA, namely 5-methylcytosine (5mC), 5-hydroxymethylcytosine (5hmC), 5-formylcytosine (5fC), and 5-carboxylcytosine (5caC). While 5mC, 5hmC, and 5fC occur in embryonic stem (ES) cells and many cell types in all the major organs (e.g. brain, heart, lung and kidney), 5caC has only been detected in a few cell types at a 10 times lower level than 5fC. The sequential oxidation of 5mC to 5hmC, 5fC, and 5caC catalyzed by the ten-eleven translocation (TET) enzymes has been proposed as a potential route for the demethylation of 5mC back to cytosine (C). Thymine DNA glycosylase (TDG), for example, can excise 5fC and 5caC from DNA as part of the base excision repair (BER) pathway to restore the C-nucleotide. Emerging evidences suggest that the different modifications may be selectively recognized by various proteins, and methyl-CpG-binding protein 2 (MeCP2) binding to 5hmC has been proposed as a means of epigenetic regulation in neurons. The conversion of 5hmC to 5fC, in turn, induces an enhanced binding of DNA-repair-associated proteins to 5fC, which has been proposed as a signal for DNA demethylation.

Specificity

Clone EDL MC-4 targets 5-methylcytosine (5mC), but not 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine (C). A less than 25% cross-reactivity toward 5-formylcytosine (5fC) was observed, which has no impact on 5mC detection by clone EDL MC-4 in cell samples, where 5mC modification level is typically more than 100- to 1,000-fold of 5fC modification level.
Target modification is not species-specific

Immunogen

BSA-conjugated 5-methylcytosine (5mC).

Application

Anti-5-Methylcytosine (5mC), clone EDL MC-4, Cat. No. MABE1081, is a highly specific mouse monoclonal antibody that targets DNA with 5-methylcytosine (5mC) modification and has been tested in Dot Blot and Immunoprecipitation.
Dot Blot Analysis: 0.5 µg/mL from a representative lot detected 0.39-100 ng of DNA with 5-methylcytosine (5mC) modification, but not DNA with only 5-hydroxymethylcytosine (5hmC) modification or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).

Immunoprecipitation Analysis: 2.5 µg from a representative lot immunoprecipitated 5 ng of DNA with 5-methylcytosine (5mC) modification, but not DNA with only 5-hydroxymethylcytosine (5hmC) modification or unmodified cytosine (Courtesy of Jeong-Heon Lee, Ph.D.; Chad Clark; Tamas Ordog, M.D.; Zhiguo Zhang, Ph.D. Epigenomics Development Laboratory, Epigenomics Program, Center for Individualized Medicine, Mayo Clinic, Rochester, Minnesota, USA).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Dot Blot analysis of DNA cytosine modifications.

Dot Blot Analysis: 2 µg/mL of this antibody detected 25-100 ng of DNA with 5-methylcytosine (5mC), exhibiting less than 25% cross-reactivity toward 5-formylcytosine (5fC) and no detectable immunoreactivity toward 5-hydroxymethylcytosine (5hmC), 5-carboxylcytosine (5caC), or unmodified cytosine.

Target description

Variable, depending on the size(s) of the DNA molecule(s) with 5-methylcytosine (5mC) modification.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Jia et al.
BMC biotechnology, 18(1), 7-7 (2018-02-08)
Detection of DNA methylome at single-base resolution is a significant challenge but promises to shed considerable light on human disease etiology. Current technologies could not detect DNA methylation genome-wide at single-base resolution with small amount of sequencing data and could
Jujun Zhou et al.
The Journal of biological chemistry, 299(8), 105017-105017 (2023-07-07)
Much is known about the generation, removal, and roles of 5-methylcytosine (5mC) in eukaryote DNA, and there is a growing body of evidence regarding N6-methyladenine, but very little is known about N4-methylcytosine (4mC) in the DNA of eukaryotes. The gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service