Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunoprecipitation (IP): 2-4 μg/mg, western blot: 1:2000-1:10,000
accession no.
NP_001370.1
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... DNMT1(1786)
Immunogen
The epitope recognized by PLA0011 maps to a region between residue 700 and 750 of human DNA-Methyltranserase 1 using the numbering given in entry NP_001370.1 (GeneID 1786).
Physical form
Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide
Other Notes
DNA methyltransferase 1 (DNMT1) catalyzes the methylation of the 5′-cytosine in the dinucleotide sequence, CpG. DNMT1 is thought to be responsible for the regulation of methylation patterns that are important in development and influence gene expression in a time- and tissue-specific manner.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.