Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE1031

Sigma-Aldrich

Anti-poly-ADP-ribose binding reagent

Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.

Synonym(s):

poly-ADP-ribose binding reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160405
NACRES:
NA.41

biological source

Escherichia coli

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

species reactivity

human, mouse

species reactivity (predicted by homology)

all

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

General description

Cat. No. MABE1031, Anti-poly-ADP-ribose binding reagent, is a His-tagged recombinant protein fused to rabbit Fc tag, expressed in and purified from Rosetta(DE3)pLysS strain of E. coli (Cat. No. 70956). Anti-poly-ADP-ribose binding reagent is useful for the affinity detection of oligo- and poly-ADP-ribosylated (PARylated) proteins on membranes or on fixed cells in a manner similar to antibody-based Western blot, dot blot, and immunocytochemistry applications. The rabbit Fc tag allows visualization of the binding/labeling with conjugated anti-rabbit secondary antibodies. The Fc tag also allows Anti-poly-ADP-ribose binding reagent to be captured on Protein A resins for affinity pull-down applications.

Specificity

Two or more units of ADP-ribose

Application

Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.
Dot Blot Specificity Analysis: This reagent detected oligo(ADPR) and poly(ADPR) on ADP-ribosylated PARP1 recombinant protein (Lee Kraus, University of Texas Southwestern Medical Center).
Immunocytochemistry Analysis: A representative lot detected oligo(ADPR) and poly(ADPR)/PAR in 3T3-L1 cells (Lee Kraus, University of Texas Southwestern Medical Center).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
General Post-translation Modification

Quality

Evaluated by Western Blotting on ADP-ribosylated PARP1 and PARP3 recombinant proteins.

Western Blotting Analysis: This reagent detected oligo(ADPR) and poly(ADPR) on ADP-ribosylated PARP1 recombinant protein (Lee Kraus, University of Texas Southwestern Medical Center).

Target description

Variable depending on the target proteins and the extend of ADP-ribosylation

Physical form

Format: Purified
Ni-NTA agarose
Purified from E. coli by Ni-NTA agarose. Supplied in buffer containing 10 mM Tris pH 7.5, 0.2 M NaCl, 10% Glycerol, 10 mM Imidazole, 1 mM PMSF, 1 mM β-Mercaptoethanol, 10% glycerol without preservatives.

Storage and Stability

Stable for 1 year at -80°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -80°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Luo et al.
Molecular cell, 65(2), 260-271 (2017-01-21)
Poly(ADP-ribosyl)ation (PARylation) is a post-translational modification of proteins mediated by PARP family members, such as PARP-1. Although PARylation has been studied extensively, few examples of definitive biological roles for site-specific PARylation have been reported. Here we show that C/EBPβ, a key
George E Ronson et al.
Nature communications, 9(1), 746-746 (2018-02-23)
PARP1 regulates the repair of DNA single-strand breaks generated directly, or during base excision repair (BER). However, the role of PARP2 in these and other repair mechanisms is unknown. Here, we report a requirement for PARP2 in stabilising replication forks
Anna-Lena Kolb et al.
Methods in molecular biology (Clifton, N.J.), 1813, 125-148 (2018-08-12)
The amoeba Dictyostelium discoideum is a single-cell organism that can undergo a simple developmental program, making it an excellent model to study the molecular mechanisms of cell motility, signal transduction, and cell-type differentiation. A variety of human genes that are
Lesley B Conrad et al.
Molecular cancer therapeutics, 19(1), 282-291 (2019-10-09)
Inhibitors of nuclear PARP enzymes (e.g., PARP-1) have improved clinical outcomes in ovarian cancer, especially in patients with BRCA1/2 gene mutations or additional homologous recombination (HR) DNA repair pathway deficiencies. These defects serve as biomarkers for response to PARP inhibitors
Tom P Aird et al.
American journal of physiology. Endocrinology and metabolism, 321(6), E802-E820 (2021-11-09)
Sprint interval training (SIT) is a time-efficient alternative to endurance exercise, conferring beneficial skeletal muscle metabolic adaptations. Current literature has investigated the nutritional regulation of acute and chronic exercise-induced metabolic adaptations in muscle following endurance exercise, principally comparing the impact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service