Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AM80

Sigma-Aldrich

Anti-PAR (Ab-1) Mouse mAb (10H)

liquid, ≥95% (SDS-PAGE), clone 10H

Synonym(s):

Anti-Poly (ADP-Ribose)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

10H, monoclonal

Assay

≥95% (SDS-PAGE)

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, Drosophila, rat, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG3

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Protein A purified mouse monoclonal antibody. Recognizes poly-ADP-ribose.
Recognizes poly(ADP)-ribose (PAR) in etopside-treated HL-60 cells.
This Anti-PAR (Ab-1) Mouse mAb (10H) is validated for use in Immunoblotting, Immunocytochemistry for the detection of PAR (Ab-1).

Immunogen

Bovine
purified poly(ADP-ribose)

Application

Immunoblotting (2.5 µg/ml)

Immunocytochemistry (5-20 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 100 mM NaCl, 50 mM HEPES, 1% BSA, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Negative Control
Normal mouse IgG
Positive Control
Etoposide-treated HL-60 cells

Other Notes

Kawamitsu, H., et al. 1984. Biochemistry23, 3771.
PAR can be induced in HL-60 cells by treatment with 67 µM etoposide for 3.5 h. Antibody should be titrated for optimal results in an individual system.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gautier Stoll et al.
Oncoimmunology, 8(6), e1571389-e1571389 (2019-05-10)
The expression of two metabolic enzymes, i.e., aldehyde dehydrogenase 7 family, member A1 (ALDH7A1) and lipase C, hepatic type (LIPC) by malignant cells, has been measured by immunohistochemical methods in non-small cell lung carcinoma (NSCLC) biopsies, and has been attributed
Laura Maggi et al.
European journal of immunology, 40(8), 2174-2181 (2010-05-21)
We have previously shown that human Th17 lymphocytes are characterized by the selective expression of IL-23 receptor (IL-23R), CCR6, CD161, and the transcription factor retinoic acid-related orphan receptor C (RORC), and originate from a CD161(+)CD4(+) naïve T-cell precursor in response
Young-Ok Son et al.
Toxicology and applied pharmacology, 255(3), 287-296 (2011-07-20)
Cadmium is a toxic heavy metal which is environmentally and occupationally relevant. The mechanisms underlying cadmium-induced autophagy are not yet completely understood. The present study shows that cadmium induces autophagy, as demonstrated by the increase of LC3-II formation and the
Li-Li Zhuang et al.
PloS one, 8(10), e77304-e77304 (2013-11-10)
All-trans retinoic acid (ATRA) is an active metabolite of Vitamin A, it shows protective effects on asthma, including maintains airway epithelial integrity, inhibits asthma effector cells differentiation, modulates immune response, et al. However, the promoting effect of ATRA on Th2
Lingyu Zhang et al.
ACS omega, 7(3), 2571-2582 (2022-02-01)
Etoposide (VP-16) is used for the treatment of various cancers, including nasopharyngeal carcinoma (NPC); however, cancers develop resistance to this agent by promoting DNA repair. The DNA-PK (DNA-PKcs) catalytic subunit and poly(ADP-ribose) polymerase 1 (PARP1) mediate acquired resistance and poor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service