Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABC684

Sigma-Aldrich

Anti-SOD1 Antibody, clone 6F5

ascites fluid, clone 6F5, from mouse

Synonym(s):

ALS, SOD, ALS1, IPOA, homodimerSuperoxide dismutase [Cu-Zn], Superoxide dismutase 1, hSod1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

6F5, monoclonal

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOD1(6647)

General description

Superoxide dismutase 1 (SOD1) also known as Superoxide dismutase [Cu-Zn] to distinguish it from SOD2 which uses manganese as its cofactor, is the cytosolic form of the enzyme that catalyzes the breakdown of superoxide into oxygen and hydrogen peroxide and is our primary anti-oxidant combating enzyme. SOD1 performs its function with the help of co-factors and one protein in particular is vital to its proper functioning. Copper chaperone protein (CCS) is a metalloprotein that specifically delivers Cu to SOD1 and may activate SOD1 through direct insertion of the Cu cofactor. Interestingly, defects in SOD1 are the cause of a form of ALS (Amyotrophic Lateral Sclerosis or Lou Gehrig′s disease) and much research is centered on understanding how over-active or under-active SOD1 contributes to the disease. EMD-Millipore′s Anti-SOD1 mouse monoclonal antibody has been tested in western blot on HeLa, NIH3T3, A549 and A431 cell lysates and fluorescent immunocytochemistry on PANC1, SKBR-3 and 3T3-L1 cells in culture. The antibody has also been tested in flow cytometry on A431 cells in culture.

Immunogen

Purified recombinant fragment of human SOD1 expressed in E. Coli.

Application

Anti-SOD1 Antibody, clone 6F5 is a highly specific mouse monoclonal antibody, that targets Superoxide Dismutase & has been tested in western blotting, ICC & Flow Cytometry.
Immunocytochemistry Analysis: A 1:200-1,000 dilution from a representative lot detected SOD1 in PANC-1, SKBR-3, and 3T3-L1 cells.

Flow Cytometry Analysis: A 1:200-400 dilution from a representative lot detected SOD1 in non serum starved A431 cells.

Optimal working dilutions must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in HeLa, NIH/3T3, A549, and A431 cell lysates.

Western Blotting Analysis: A 1:500-2,000 dilution of this antibody detected SOD1 in HeLa, NIH/3T3, A549, and A431 cell lysates.

Target description

~18 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
HeLa, NIH/3T3, A549, and A431 cell lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Dan Hao et al.
Investigative ophthalmology & visual science, 63(13), 1-1 (2022-12-02)
The purpose of this study was to identify a new candidate gene for keratoconus and congenital cataracts and further investigate its underlying pathogenic mechanisms. This study, using a Chinese family with keratoconus and congenital cataracts, 262 patients with sporadic keratoconus
Xiao-Dan Hao et al.
Scientific reports, 7(1), 13570-13570 (2017-10-21)
Keratoconus (KC) is a common degenerative corneal disease, and heredity plays a key role in its development. Although few genes are known to cause KC, a large proportion of disease-causing genes remain to be revealed. Here, we report the identification
Joana V Silva et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Aging is associated with testicular morphological and functional alterations, but the underlying molecular mechanisms and the impact of physical exercise are poorly understood. In this study, we examined the effects of age and lifelong moderate-intensity exercise on rat testis. Mature
Glyn Chidlow et al.
Frontiers in neuroscience, 11, 478-478 (2017-09-09)
The vascular hypothesis of glaucoma proposes that retinal ganglion cell axons traversing the optic nerve head (ONH) undergo oxygen and nutrient insufficiency as a result of compromised local blood flow, ultimately leading to their degeneration. To date, evidence for the
Adrián Jordá et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Ranolazine (Rn) is a drug used to treat persistent chronic coronary ischemia. It has also been shown to have therapeutic benefits on the central nervous system and an anti-diabetic effect by lowering blood glucose levels; however, no effects of Rn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service