Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABC539

Sigma-Aldrich

Anti-SFRP2 Antibody, clone 80.8.6, azide free

clone 80.8.6, from mouse

Synonym(s):

Secreted frizzled-related protein 2, FRP-2, sFRP-2, Secreted apoptosis-related protein 1, SARP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

80.8.6, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable
inhibition assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SFRP2(6423)

General description

Secreted frizzled-related proteins (SFRPs) may play a critical role in carcinogenesis by inhibiting the Wnt/beta-catenin signaling pathway. Secreted frizzled-related protein 2 (SFRP2) may be useful to further investigate mammary tumorigenesis as its expression increases with tumor progression in canine mammary gland tumors. SFRP2 in mouse embryos has been found to cause severe shortening of the thoracic region. Studies have shown that this protein is the essential stem cell paracrine factor that is thought to regulate myocardial survival and repair damage in the aftermath of an ischemic injury.

Immunogen

Epitope: NTR domain
Linear peptide corresponding to the NTR domain of human SFRP2.

Application

Anti-SFRP2 Antibody, clone 80.8.6, azide free is an antibody against SFRP2 for use in IHC & inhibition.
Inhibition Assay Analysis: Anti-SFRP2, clone 80.8.6 inhibits tube formation and cell migration in vitro on SVR angiosarcoma cells, inhibits angiosarcoma and breast cancer growth in vivo, and increases nuclear NFATc3 levels.

Quality

Evaluated by Immunohistochemistry in human prostate and human breast cancer tissue.

Imunnohistochemistry Analysis: A 1:500 dilution of this antibody detected SFRP2 in human prostate and human breast cancer tissue.

Target description

33 kDa calculated

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanpreet Kaur et al.
Nature, 532(7598), 250-254 (2016-04-05)
Cancer is a disease of ageing. Clinically, aged cancer patients tend to have a poorer prognosis than young. This may be due to accumulated cellular damage, decreases in adaptive immunity, and chronic inflammation. However, the effects of the aged microenvironment
Mitchell E Fane et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(21), 5709-5719 (2020-10-25)
Angiogenesis is thought to be critical for tumor metastasis. However, inhibiting angiogenesis using antibodies such as bevacizumab (Avastin), has had little impact on melanoma patient survival. We have demonstrated that both angiogenesis and metastasis are increased in older individuals, and
Y Sun et al.
Oncogene, 35(33), 4321-4334 (2016-01-12)
Most tumors initially respond to cytotoxic treatments, but acquired resistance often follows. The tumor microenvironment (TME) is a major barrier to clinical success by compromising therapeutic efficacy, and pathological relevance of multiple soluble factors released by a therapeutically remodeled TME

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service