Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB9670

Sigma-Aldrich

Anti-Echovirus Blend Antibody, blend of sixs

ascites fluid, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

Specificity

Reacts with the Echovirus 4, 6, 9, 11, 30 and 34.

CROSS REACTIVITIES:

No cross-reactivity to CMV, HSV 1 & 2, Para 1, 2, & 3, Poliovirus 1 & 3 or Mumps.

Immunogen

Epitope: blend of six monoclonals

Application

Anti-Echovirus Blend Antibody, blend of sixs is an antibody against Echovirus Blend for use in IF.
IFA at 1:100.

Dilute with buffer pH 7.5-8.0 containing protein or other stabilizing medium.

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Ascites, with 0.1% sodium azide.

Storage and Stability

Maintain at -20*C in convenient, undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Luo et al.
Frontiers in cellular and infection microbiology, 14, 1391824-1391824 (2024-07-24)
E11 causes acute fulminant hepatitis in newborns. We investigated the pathological changes of different tissues from premature male twins who died due to E11 infection. The E11 expression level was higher in the liver than in other tissues. IP10 was
Kuan-Chi Tseng et al.
Viruses, 14(1) (2022-01-23)
Enterovirus 71 (EV71) is an etiological agent of hand foot and mouth disease and can also cause neurological complications in young children. However, there are no approved drugs as of yet to treat EV71 infections. In this study, we conducted
Hsin-Yu Hou et al.
The Journal of general virology, 97(5), 1122-1133 (2016-02-18)
Enterovirus 71 (EV71) causes life-threatening diseases with neurological manifestations in young children. However, the treatment of EV71 infections remains an unmet medical need. Idarubicin (IDR) is an anthracycline compound that is used therapeutically for certain types of tumour. In this
Yu-Siang Su et al.
Frontiers in microbiology, 11, 1656-1656 (2020-08-08)
Enterovirus A71 (EV-A71) is one of the major etiologic agents causing hand, foot, and mouth disease (HFMD) in children and occasionally causes severe neurological diseases or even death. EV-A71 replicates rapidly in host cells. For a successful infection, viruses produce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service