Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB8262

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H

clone 133-1H, Chemicon®, from mouse

Synonym(s):

RSV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

133-1H, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG2aκ

shipped in

wet ice

Specificity

Directed against the 47-49 kDa Fusion Protein of RSV. Antibody MAB858-1 recognizes both A and B RSV strains.

Immunogen

Respiratory Syncytial virus A2

Application

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H is an antibody against Respiratory Syncytial Virus for use in ELISA & IF.
ELISA, Indirect Immunofluorescence

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Format: Purified
Purified. Liquid 0.02M PBS, 0.25M NaCl, 0.1% sodium azide

Storage and Stability

Store at 2 to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A highly stable prefusion RSV F vaccine derived from structural analysis of the fusion mechanism.
Krarup, A; Truan, D; Furmanova-Hollenstein, P; Bogaert, L; Bouchier, P; Bisschop et al.
Nature Communications null
Respiratory syncytial virus NS1 protein colocalizes with mitochondrial antiviral signaling protein MAVS following infection.
Boyapalle, S; Wong, T; Garay, J; Teng, M; San Juan-Vergara, H; Mohapatra, S; Mohapatra, S
Testing null
Bárbara N Porto et al.
Viruses, 13(10) (2021-10-27)
Respiratory Syncytial Virus (RSV) is the leading cause of acute lower respiratory infections in young children and infection has been linked to the development of persistent lung disease in the form of wheezing and asthma. Despite substantial research efforts, there
Catherine A A Beauchemin et al.
PloS one, 14(4), e0214708-e0214708 (2019-04-16)
Many aspects of the respiratory syncytial virus (RSV) are still poorly understood. Yet these knowledge gaps have had and could continue to have adverse, unintended consequences for the efficacy and safety of antivirals and vaccines developed against RSV. Mathematical modelling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service