Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MABX34

Sigma-Aldrich

Anti-Respiratory Syncytial Virus (RSV)

Synonym(s):

Anti-RSV fusion protein Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX34-1KC)
pkg of 10 mg (MABX34-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable
immunofluorescence: suitable

isotype

IgG1

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

Fusion protein of RSV types A & B

Immunogen

Native RSV

Application

This mouse monoclonal antibody, Cat. No. MABX34, is tested for use in Immunofluorescence, Lateral Flow Assay, and ELISA, for the detection of Fusion protein of RSV strains A & B.

Target description

Respiratory syncytial virus (RSV) is a common respiratory virus that usually causes mild, cold-like symptoms. Most people recover in a week or two, but RSV can be serious, especially for infants and older adults.

Linkage

Recommended Matched Pair MABX34 capture and MABX35 detection / MABX35 can self-pair capture and detection.

Physical form

Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.2 with 0.1% sodium azide.

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service