Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB1964-I

Sigma-Aldrich

Anti-Integrin β4 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA
ICC
IHC (p)
WB
clone:
3E1, monoclonal
species reactivity:
human
citations:
technique(s):
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3E1, monoclonal

mol wt

calculated mol wt 202.17 kDa

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB4(3691)

General description

Integrin β4 (UniProt: P16144; also known as GP150, CD104) is encoded by the ITGB4 gene (Gene ID: 3691) in human. Integrins are heterodimeric cell surface receptors composed of a variable alpha subunit and a conserved beta subunit. The exact combination of various alpha and beta subunits dictates the binding specificity of integrins to different ECM components. Integrins are not constitutively active. Activation of an integrin from low ligand-binding affinity state to high ligand-binding affinity state requires conformational change that can originate from either from their cytoplasmic or extracellular interactions. The recognition site for most integrins that bind the ECM consists of an RGD (arginine-glycine-aspartic acid) sequence. Integrins bind to their ligands with low affinity and this binding occurs only when a certain minimum number of integrins are present at specific points known as focal contacts. Integrin β4 is a single-pass type I membrane protein that is synthesized with a signal peptide (aa 1-27), which is subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 28-710), a transmembrane domain (aa 711-733), and a cytoplasmic domain (aa 734-1822). It contains four fibronectin type III domains. Integrin β4 heterodimerizes with a6 that serves as a receptor for laminin and plays a critical structural role in the hemidesmosome of epithelial cells. It undergoes palmitoylation by DHHC3 at several cysteines of the membrane-proximal region that enhances its stability and surface expression. Palmitoylation also promotes its secondary association with tetraspanins.

Specificity

Clone 3E1 is a mouse monoclonal antibody that detects Integrin β4. It targets an epitope within the extracellular domain.

Immunogen

Human fetal cell membranes.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: A 1:1,000 dilution of this antibody detected Integrin β4 in A431 cells.

Tested Applications

Flow Cytometry Analysis: 1 βg from a representative lot detected Integrin β4 in one million A431 cells.

Flow Cytometry Analysis: A representative lot detected Integrin β4 in Flow Cytometry applications (Vijayakumar, S., et. al. (2008). J. Am. Soc. Nephrol.19(6):1079-91; Hauptmann, S., et. al. (1995). Int. J. Cancer. 61(6):819-25).

Immunohistochemistry Applications: A representative lot detected Integrin β4 in Immunohistochemistry applications ( Hauptmann, S., et. al. (1995). Int. J. Cancer. 61(6):819-25).

Immunofluorescence Analysis: A representative lot detected Integrin β4 in Immunofluorescence applications (Swanner, J., et. al. (2019). FASEB BioAdv. 1(10):639-660).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Integrin β4, clone 3E1, Cat. No. MAB1964-I, is a mouse monoclonal antibody that detects Integrin β4 and is tested for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at +2°C to +8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service