Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

MABE952

Sigma-Aldrich

Anti-Histone H3.1 Antibody

rat monoclonal, 1D4F2

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Histone H3.1 Antibody, clone 1D4F2, clone 1D4F2, 1 mg/mL, from rat

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D4F2, monoclonal

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

ChIP: suitable (ChIP-seq)
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

KLH-conjugated linear peptide corresponding to human Histone H3.1.

Application

Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected Histone H3.1 in HeLa cells.

Immunocytochemistry Analysis: A 1:2,000 dilution from a representative lot detected Histone H3.1 in NIH/3T3 cells (Prof. Taro Tachibana, Cell Engineering Corporation.).

Alexa Fluor is a registered trademark of Life Technologies.

Immunoprecipitation Analysis: A representative lot from an independent laboratory detected Histone H3.1 in C2C12 cells (Harada, A., et al. (2012). EMBO J. 31(13):2994-3007.).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Histone H3.1 (Maehara, K., et al. (2013). Nucleic Acids Res. 41(1):54-62.).

Chromatin Immunoprecipitation Sequencing Analysis: A representative lot from an independent laboratory immunoprecipitated Histone H3.1 (Maehara, K., et al. (2013). Nucleic Acids Res. 41(1):54-62.).
This Anti-Histone H3.1 Antibody, clone 1D4F2 is validated for use in western blotting, ICC, IP, ChIP & ChIP-seq for the detection of Histone H3.1.

Quality

Evaluated by Western Blotting in HeLa acid extract.

Western Blotting Analysis: 1 µg/mL of this antibody detected Histone H3.1 in 10 µg of HeLa acid extract.

Target description

~17 kDa observed

Physical form

Format: Purified

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service