Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB19101

Sigma-Aldrich

Anti-Tenascin Antibody, clone DB7

clone DB7, Chemicon®, from mouse

Synonym(s):

Tenascin-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

DB7, monoclonal

species reactivity

human, rabbit, guinea pig

should not react with

rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TNC(3371)

Specificity

The antibody recognizes the Mr 250000 and 180000 tenascin polypeptides in immunoprecipitation and immunoblots the Mr 250000 polypeptide. The antibody reacts with the fibrinogen-like knob-domain of tenascin protein

Immunogen

Tenascin polypeptides isolated from spent culture supernatant of human fibroblasts isolated by affinity chromatography.

Application

Detect Tenascin using this Anti-Tenascin Antibody, clone DB7 validated for use in IF, WB, IH(P).
Immunofluorescence: 1-2% PFA fixed, permeabilized tissues or cell cultures. Mild pepsin digestion improves staining.

Immunohistochemistry (formalin-fixed and paraffin-embedded sections): 1:200-1:400.

Requires pepsin digestion (0.1-.4% pepsin in .1% HCL pH 1.0, 30-50 minutes at 37C; neutralize sections with 2 quick rinses in 1X PBS pH 7.4 prior to blocking and antibody staining. Enhanced ABC or amplified detection systems recommended for best signals. See Soini, Y 1993 J Clin Patholo for protocol for decalified bone.

Immunoblotting: 1:1000; SDS sample buffer lysis of whole human fibroblasts; 6.5% PAGE reduced gels. Two distinct bands, 250,000 & 190,000 {Soini, Y Am. J. Clin. Path. 1993)

Final dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Linkage

Replaces: MAB1902

Physical form

Format: Purified
The purified antibody is in PBS solution containing 1.0% (w/v) BSA and 0.1% (w/v) sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefanie Deinhardt-Emmer et al.
eLife, 10 (2021-03-31)
Clinical observations indicate that COVID-19 is a systemic disease. An investigation of the viral distribution within the human body and its correlation with tissue damage can aid in understanding the pathophysiology of SARS-CoV-2 infection. We present a detailed mapping of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service