Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT879

Sigma-Aldrich

Anti-Thrombospondin-1 Antibody, clone 133

clone 133, from mouse

Synonym(s):

Thrombospondin-1, THBS-1, TSP-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

133, monoclonal

species reactivity

rat, human

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
neutralization: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... THBS1(7057)

General description

Thrombospondin-1 (UniProt P07996; also known as THBS-1, TSP-1) is encoded by the THBS1 (also known as THBS, TSP, TSP1) gene (Gene ID 7057) in human. Thrombospondin-1 (TSP-1) is a member of the TSP family of calcium-binding extracellular matrix proteins. The TSP family consists of the homotrimeric TSP-1 and TSP-2, as well as the homopentameric TSP-3, TSP-4 and TSP-5/COMP (cartilage oligomeric matrix protein). TSP-1 is induced at injury site and functions as an activator of latent TGF-β. TSP-1 binding alters the conformation of the latent TGF-β complex and renders TGF-β biologically active, which in turn can also induce TSP-1 expression. TSP-1 mediates the proliferation of fibroblasts and smooth muscle cells, while it inhibits the proliferation of endothelial cells. TSP-1 may also serve as both an attachment protein and an anti-adhesive molecule as shown by its ability to cause disassembly of focal adhesions of endothelial cells. TSP-1 is initially produced with a signal peptide sequence (a.a. 1-18), the removal of which yields the mature protein (a.a. 19-1170). Structurally, TSP-1 monomer consists of a N-terminal heparin-binding (a.a. 47-95) region and a laminin G-like domain (a.a. 65-270), followed by an oligomerization domain (a.a. 259-311), a procollagen or VWFC domain (a.a. 316-373), three properdin or type I repeats (a.a. 379-429, 435-490, and 492-547), two EGF-like domains (a.a. 547-587 and 646-690), eight type III repeats or calcium-wire module (a.a. 691-954), and a C-terminal lectin-like globular module or G domain (a.a. 958-170).

Specificity

Clone 133 reacted with human thrombospondin-1 (hTSP-1), but not hTSP-2, mouse TSP-1 (mTSP-1), or mTSP-2. Clone 133 detected the C-terminal 50 kDa chymotryptic fragment of stripped TSP as well as recominant fragements containing the Calcium-binding type-3 repeats (a.a. 692-945) region (Annis, D.S., et al. (2006). J. Thromb. Haemost. 4(2):459-468; Schultz-Cherry, S., and Murphy-Ullrich J.E. (1993). J. Cell Biol. 122(4):923-932).

Immunogen

Alkaline-stripped human platelet thrombospondin-1 (sTSP-1).
Epitope: Calcium-binding type-3 repeats.

Application

Detect Thrombospondin-1 using this Anti-Thrombospondin-1 Antibody, clone 133 validated for use in Western Blotting, Immunohistochemistry (Paraffin), Neutralizing, ELISA.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western Blotting Analysis: A representative lot detected an upregulated thrombospondin-1 (TSP-1) expression in cultured primary rat vascular smooth muscle cells (VSMCs) upon treatment with either stainless steel (SS) ions or active TGF-β (Pallero, M.A., et al. (2010). J. Vasc. Res. 47(4):309-322).
Western Blotting Analysis: A representativev lot ditected full-length human thrombospondin-1 (hTSP-1) as well as hTSP-1 C-terminal fragments E3CaG (a.a. 648-1170), E3Ca (a.a. 648-945) and Ca (a.a. 692-945), but not hTSP-2, mouse TSP-1 (mTSP-1), or mTSP-2 by Western blotting under either reducing or non-reducing condition (Annis, D.S., et al. (2006). J. Thromb. Haemost. 4(2):459-468).
Western Blotting Analysis: A representative lot detected an upregulated thrombospondin-1 (TSP-1) in human pancreatic cancer Panc-1 cells upon siRNA-mediatted k-Ras knockdown (Fleming, J.B., et al. (2005). Mol. Cancer Res. 3(7):413-423).
Immunohistochemistry Analysis: A representative lot detected elevated thrombospondin-1 (TSP-1) immunoreactivity in vascular tissues surrounding stent materials due to in-stent restenosis (ISR) in paraffin-embedded sections of human coronary arteries from patients received either Taxus or Cypher drug-eluting stent implant (Pallero, M.A., et al. (2010). J. Vasc. Res. 47(4):309-322).
Neutralizing Analysis: Representative lots prevented alkaline-stripped human platelet thrombospondin-1 (sTSP-1) from activatating the latent form of TGF-beta secreted by cultured NMuMG murine mammary gland epithelial cells or bovine aortic endothelial (BAE) cells without affecting cellular signaling induced by the mature/active form of TGF-beta (Alcaraz, L.B., et al. (2014). J. Cell Biol. 205(3):409-428; Annis, D.S., et al. (2006). J. Thromb. Haemost. 4(2):459-468; Schultz-Cherry, S., and Murphy-Ullrich J.E. (1993). J. Cell Biol. 122(4):923-932).
Neutralizing Analysis: A representative lot inhibited stainless steel (SS) ions-induced TGF-β signaling in cultured primary rat vascular smooth muscle cells (VSMCs) as indicated by a suppressed induction of the synthetic/myofibroblastic protein ED-A FN (Pallero, M.A., et al. (2010). J. Vasc. Res. 47(4):309-322).
ELISA Analysis: A representativev lot ditected full-length human thrombospondin-1 (hTSP-1) as well as hTSP-1 C-terminal fragments E3CaG (a.a. 648-1170) and E3Ca (a.a. 648-945), but not hTSP-2, mouse TSP-1 (mTSP-1), or mTSP-2 by direct/non-sandwich ELISA (Annis, D.S., et al. (2006). J. Thromb. Haemost. 4(2):459-468).

Quality

Evaluated by Western Blotting in human thrombospondin-1 purified protein.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected Thrombospondin-1 in 0.1 µg of human thrombospondin-1 purified protein.

Target description

~129 kDa observed.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lactic acidosis induces metabolic and phenotypic reprogramming in cholangiocarcinoma cells via the upregulation of thrombospondin-1.
Thamrongwaranggoon, et al.
Cancer Science, 114, 1541-1555 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service