Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAB10224

Sigma-Aldrich

Anti-L. pneumophila Serogroup 1 Antibody, clone Lp1 3

ascites fluid, clone Lp1 MAB 3, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

Lp1 MAB 3, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2b

shipped in

dry ice

General description

Legionella pneumophila are Gram-negative bacteria which are associated with Legionnaires′ disease and Pontiac fever. At least 14 different serovars of L. pneumophila have been described as well as several other species being subdivided into a number of serovars.

Specificity

The antibody reacts with a surface antigen on 2 of the 10 antigenic subgroups of L. pneumophila

Immunogen

Bacterial antigenic subgroup Knoxville 1

Application

This Anti-L. pneumophila Serogroup 1 Antibody, clone Lp1 3 is validated for use in IF for the detection of L. pneumophila Serogroup 1.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J R Joly et al.
Journal of clinical microbiology, 23(4), 768-771 (1986-04-01)
A panel of monoclonal antibodies to Legionella pneumophila serogroup 1 and a subclassification scheme were developed in a collaborative project among three laboratories. The seven most useful monoclonal antibodies were selected from three previously developed panels on the basis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service