Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LX0020

Supelco

Lactic Acid

85%, meets analytical specifications of USP/NF GR ACS

Synonym(s):

2-Hydroxypropionic Acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106

Product Name

Lactic Acid 85%, meets analytical specifications of USP/NF GR ACS

grade

ACS reagent

Quality Level

Agency

meets analytical specifications of USP/NF GR ACS

form

liquid

concentration

85.0-90.0% (lactic acid)

impurities

≤0.001% Chloride (Cl)
≤0.002% Sulfate (SO4)
≤5 ppm Heavy metals (as Pb)
≤5 ppm Iron (Fe)

ign. residue

≤0.02%

color

APHA: ≤50

shipped in

ambient

storage temp.

room temp

General description

Meets ACS Specifications GR ACS

Application

Meets ACS Specifications GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay: 85.0-90.0%
Chloride (Cl): 0.001% max
Color (APHA): 50 max
Form: Liquid
Heavy metals (as Pb): 5ppm max
Iron (Fe): 5ppm max
Residue after ignition: 0.02% max
Substances darkened by sulfuric acid: To pass test
Sulfate (SO4): 0.002% max

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service